Skip to Content
Merck
All Photos(1)

Documents

444367

Sigma-Aldrich

5-Amino-2,4,6-triiodoisophthalic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6I3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
558.84
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

reaction suitability

reaction type: solution phase peptide synthesis

impurities

<5% water

mp

265-270 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1c(I)c(C(O)=O)c(I)c(C(O)=O)c1I

InChI

1S/C8H4I3NO4/c9-3-1(7(13)14)4(10)6(12)5(11)2(3)8(15)16/h12H2,(H,13,14)(H,15,16)

InChI key

JEZJSNULLBSYHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service