442658
Ethynylferrocene
97%
Synonym(s):
Ferrocenylacetylene, Ferrocenylethyne
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HC≡CC5H4)Fe(C5H5)
CAS Number:
Molecular Weight:
210.05
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
reaction suitability
core: iron
reagent type: catalyst
mp
50-54 °C (lit.)
storage temp.
2-8°C
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.C#C[C]2[CH][CH][CH][CH]2
InChI
1S/C7H5.C5H5.Fe/c1-2-7-5-3-4-6-7;1-2-4-5-3-1;/h1,3-6H;1-5H;
InChI key
HFWSJSDVVFIFRS-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gaulthier Rydzek et al.
ACS nano, 8(5), 5240-5248 (2014-04-18)
Simple preparation methods of chemically versatile and highly functionalizable surfaces remain rare and present a challenging research objective. Here, we demonstrate a simultaneous electropolymerization and electro-click functionalization process (SEEC) for one-pot self-construction of aniline- and naphthalene-based functional polymer films where
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service