440221
Vinyltrimethoxysilane
97%
Synonym(s):
(Trimethoxysilyl)ethylene, Ethenyltrimethoxysilane, Trimethoxy(vinyl)silane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2C=CHSi(OCH3)3
CAS Number:
Molecular Weight:
148.23
Beilstein:
1099136
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.392 (lit.)
bp
123 °C (lit.)
density
0.968 g/mL at 25 °C (lit.)
SMILES string
CO[Si](OC)(OC)C=C
InChI
1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3
InChI key
NKSJNEHGWDZZQF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Vinyltrimethoxysilane (VTMS) is a silane coupling agent with silicon and hydroxyl groups that can be used to enhance the wettability and improve the superhydrophobic characteristics of different composites.
Application
VTMS may be used to provide superhydrophobicity to different materials like TiO2, talc, kaolin, magnesium oxide nanoparticles, ammonium phosphate, and PEDOT. It modifies the surface by capping the material and creates a protective layer that is water resistant and can be used in major coating industries.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Flam. Liq. 2 - Skin Sens. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
71.6 °F - closed cup
Flash Point(C)
22 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and characterization of PEDOT: P (SS-co-VTMS) with hydrophobic properties and excellent thermal stability
Cho W, et al.
Polymers, 8(5), 189-189 (2016)
UV-induced switching behavior of novel fluoroalkyl end-capped vinyltrimethoxysilane oligomer/titanium oxide nanocomposite between superhydrophobicity and superhydrophilicity with good oleophobicity.
Sawada E, et al.
Composites Part B: Engineering, 41(6), 498-502 (2010)
Surface modification of ammonium polyphosphate with vinyltrimethoxysilane: Preparation, characterization, and its flame retardancy in polypropylene.
Qin Z, et al.
Polymer Degradation and Stability, 119, 139-150 (2015)
Preparation of morphology-controlled fluoroalkyl end-capped vinyltrimethoxysilane oligomeric silica/magnesium oxide nanocomposite particles: development of magnesium oxide nanocomposite particles possessing a water-resistance ability.
Oikawa Y, et al.
Journal of Sol-Gel Science and Technology, 7(25), 1-13 (2018)
Preparation and surface property of fluoroalkyl end-capped vinyltrimethoxysilane oligomer/talc composite-encapsulated organic compounds: application for the separation of oil and water.
Oikawa Y, et al.
ACS Applied Materials & Interfaces, 7(25), 13782-13793 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service