Skip to Content
Merck
All Photos(1)

Key Documents

439681

Sigma-Aldrich

Sodium iodide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Sodium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
beads

vapor density

>1 (vs air)

Quality Level

product line

AnhydroBeads

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

661 °C (lit.)

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anke de Vries et al.
Investigative radiology, 50(4), 297-304 (2015-01-01)
The objective of this study was to investigate the feasibility and the accuracy of spectral computed tomography (spectral CT) to determine the tissue concentrations and localization of high-attenuation, iodine-based contrast agents in mice. Iodine tissue concentrations determined with spectral CT
Caroline Serrano-Nascimento et al.
Molecular and cellular endocrinology, 349(2), 154-161 (2011-10-18)
Iodide excess acutely downregulates NIS mRNA expression, as already demonstrated. PCCl3 cells treated or not with NaI, NaI+NaClO(4) or NaI+Methimazole, for 30 min to 24 h, were used to further explore how iodide reduces NIS gene expression. NIS mRNA expression
Zhibo Liu et al.
Molecular pharmaceutics, 12(3), 974-982 (2015-01-30)
Bradykinin B1 receptor (B1R) is involved in pain and inflammation pathways and is upregulated in inflamed tissues and cancer. Due to its minimal expression in healthy tissues, B1R is an attractive target for the development of therapeutic agents to treat
Lan-Tao Li et al.
Chemical communications (Cambridge, England), 48(42), 5187-5189 (2012-04-20)
nBu(4)NI-catalyzed C3-selective formylation of N-H and N-substituted indoles by using N-methylaniline as a formylating reagent was first successfully demonstrated.
Jose A Castro-Osma et al.
Dalton transactions (Cambridge, England : 2003), 44(27), 12388-12400 (2014-12-24)
A series of alkyl organoaluminium complexes based on bulky heteroscorpionate ligands were designed as catalysts for the ring-opening polymerisation of cyclic esters. Thus, the treatment of AlX3 (X = Me, Et) with bulky acetamide or thioacetamide heteroscorpionate ligands nbptamH (1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service