Skip to Content
Merck
All Photos(1)

Key Documents

439053

Sigma-Aldrich

(+)-Isopulegol

99%

Synonym(s):

(1S,2R,5S)-2-Isopropenyl-5-methylcyclohexanol, (1S,3S,4R)-p-Menth-8-en-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
4659691
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D +22°, neat

refractive index

n20/D 1.469 (lit.)

bp

91 °C/12 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CC[C@@H]([C@@H](O)C1)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3/t8-,9+,10-/m0/s1

InChI key

ZYTMANIQRDEHIO-AEJSXWLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopulegol is a monoterpene alcohol, which is generally found in essential oils of various plants. It is widely used as a fragrance ingredient in cosmetics, shampoos and toilet soaps. Isopulegol is also a key intermediate in the synthesis of (−)-menthol.

Application

(+)-Isopulegol can be used as a starting material to synthesize:
  • A natural product (−)-isopiperitenone.
  • Medicinally important octahydro-2H-chromen-4-ol derivatives by reacting with benzaldehydes.
  • Antituberculosis diterpenoid named 12-epi-ileabethoxazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Central nervous system activity of acute administration of isopulegol in mice
Silva MIG, et al.
Pharmacology, Biochemistry, and Behavior, 88(2), 141-147 (2007)
Total Synthesis of Ileabethoxazole, Pseudopteroxazole, and seco-Pseudopteroxazole
Yang M, et al.
Angewandte Chemie (International ed. in English), 55(8), 2851-2855 (2016)
One-step, stereoselective synthesis of octahydrochromanes via the Prins reaction and their cannabinoid activities
Slater S, et al.
Tetrahedron Letters, 59(9), 807-810 (2018)
Fragrance material review on isopulegol
Bhatia SP, et al.
Food And Chemical Toxicology, 46(11), S185-S189 (2008)
Chemoenzymatic synthesis of the intermediates in the peppermint monoterpenoid biosynthetic pathway
Cheallaigh AN, et al.
Journal of Natural Products, 81(7), 1546-1552 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service