Skip to Content
Merck
All Photos(2)

Key Documents

437913

Sigma-Aldrich

4,4′-(9-Fluorenylidene)dianiline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N2
CAS Number:
Molecular Weight:
348.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

237-239 °C (lit.)

SMILES string

Nc1ccc(cc1)C3(c2ccc(N)cc2)c4ccccc4-c5ccccc35

InChI

1S/C25H20N2/c26-19-13-9-17(10-14-19)25(18-11-15-20(27)16-12-18)23-7-3-1-5-21(23)22-6-2-4-8-24(22)25/h1-16H,26-27H2

InChI key

KIFDSGGWDIVQGN-UHFFFAOYSA-N

General description

4,4′-(9-Fluorenylidene)dianiline is a diamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and characterization of new sulfonated polyimides as proton-exchange membranes for fuel cells.
Ye X, et al.
Journal of Membrane Science, 279(1), 570-577 (2006)
Gönül Yenilmez Çiftçi et al.
Dalton transactions (Cambridge, England : 2003), 42(41), 14916-14926 (2013-09-03)
A first series of the 4,4'-(9-fluorenylidene)diphenol (), () and 4,4'-(9-fluorenylidene)dianiline (), () bridged cyclotriphosphazene derivatives (, ) were synthesized by nucleophilic SN2(P) and SN1(P) reactions, respectively. The structural investigations of the compounds were verified by elemental analyses, mass spectrometry, UV-vis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service