Skip to Content
Merck
All Photos(2)

Documents

437239

Sigma-Aldrich

Poly(ethylene-co-vinyl acetate)

vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)m[CH2CH(OCOCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

Quality Level

autoignition temp.

500 °F

melt index

8 g/10 min (190°C/2.16kg)

contains

200-900 ppm BHT as inhibitor

composition

vinyl acetate, 18 wt. %

hardness

38 (Shore D, ASTM D 2240)

mp

87 °C

transition temp

softening point 61 °C (Vicat, ASTM D 1525)

density

0.94 g/mL at 25 °C

SMILES string

C=C.CC(=O)OC=C

InChI

1S/C4H6O2.C2H4/c1-2-3-4(5)6;1-2/h2H,1,3H2,(H,5,6);1-2H2

InChI key

DQXBYHZEEUGOBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Flexible tubing, color concentrates, gaskets and molded parts for autos, plastic lenses and pumps.

Features and Benefits

Thermoplastic, resilient, tough and resistant to ozone and environmental stress cracking. Inherently flexible, requiring no plasticizers.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Li Lee et al.
Acta biomaterialia, 8(6), 2271-2278 (2012-02-22)
Triple-layered microparticles comprising poly(D,L-lactide-co-glycolide, 50:50) (PLGA), poly(L-lactide) (PLLA) and poly(ethylene-co-vinyl acetate, 40 wt.% vinyl acetate) (EVA) were fabricated using a one-step solvent evaporation technique, with ibuprofen drug localized in the EVA core. The aim of this study was to investigate
The synthetic ethylene-vinyl acetate bench model.
Rafael Denadai et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(2), 288-289 (2012-02-09)
Wei Li Lee et al.
Journal of biomedical materials research. Part A, 100(12), 3353-3362 (2012-06-27)
Numerous models that predict drug release from nonerodible reservoir-membrane sphere systems have been presented. Most of these models cater only to a phase of drug release from a constant reservoir. All these models, however, are not applicable to drug release
Rafael Denadai et al.
Acta cirurgica brasileira, 27(3), 271-278 (2012-03-31)
To assess the acquisition of suture skills by training on ethylene-vinyl acetate bench model in novice medical students. Sixteen medical students without previous surgery experience (novices) were randomly divided into two groups. During one hour group A trained sutures on
Lili Wang et al.
PloS one, 7(6), e37781-e37781 (2012-06-14)
The study on the rare earth (RE)-doped layered double hydroxides (LDHs) has received considerable attention due to their potential applications in catalysts. However, the use of RE-doped LDHs as polymer halogen-free flame retardants was seldom investigated. Furthermore, the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service