Skip to Content
Merck
All Photos(1)

Key Documents

422231

Sigma-Aldrich

4-(Trifluoromethoxy)benzamidoxime

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4C(=NOH)NH2
CAS Number:
Molecular Weight:
220.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

113-115 °C (lit.)

functional group

amine
fluoro
oxime

SMILES string

N\C(=N/O)c1ccc(OC(F)(F)F)cc1

InChI

1S/C8H7F3N2O2/c9-8(10,11)15-6-3-1-5(2-4-6)7(12)13-14/h1-4,14H,(H2,12,13)

InChI key

COHKFOZYLCDVRK-UHFFFAOYSA-N

General description

4-(Trifluoromethoxy)benzamidoxime (4-TFMBAO) is a benzamidoxime (BAO) derivative containing amidoxime functional group. Its density and freezing point have been determined.

Application

4-(Trifluoromethoxy)benzamidoxime (4-TFMBAO) is suitable reactant in the fluorescence (FL) deriving reaction, one of the widely-used methodology specifically used to quantify uracil. It may be used as a reactant in the synthesis of oxadiazoles. It may also be used a fluorogenic agent in the quantification of orotic acid by spectrofluorometric method in human biological specimens.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 185-185 (2015)
Thomas E Barta et al.
Bioorganic & medicinal chemistry letters, 21(10), 2820-2822 (2011-04-22)
Seeking compounds preferentially potent and selective for MMP-13, we reported in the preceding Letter on a series of hydroxamic acids with a flexible benzamide tail groups.(1a) Here, we replace the amide moiety with non-hydrolyzable heterocycles in an effort to improve
Sensitive and Selective Determination of Orotic Acid in Biological Specimens Using a Novel Fluorogenic Reaction.
Yin S, et al.
Journal of Fluorescence, 25(4), 1005-1011 (2015)
Evan R Abt et al.
Cell chemical biology, 27(2), 197-205 (2019-11-18)
Biosynthesis of the pyrimidine nucleotide uridine monophosphate (UMP) is essential for cell proliferation and is achieved by the activity of convergent de novo and salvage metabolic pathways. Here we report the development and application of a cell-based metabolic modifier screening
Takayuki Shibata et al.
Analytica chimica acta, 674(2), 234-238 (2010-08-04)
Facile and specific methods to quantify a nucleobase in biological samples are of great importance for diagnosing disorders in nucleic acid metabolism. In the present study, a novel fluorogenic reaction specific for uracil has been developed. The reaction was carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service