Skip to Content
Merck
All Photos(2)

Documents

420964

Sigma-Aldrich

(1S)-(+)-Ketopinic acid

99%

Synonym(s):

(1S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptane-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein:
4180005
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]23/D +58°, c = 1 in chloroform

mp

237-239 °C (lit.)

SMILES string

CC1(C)C2CCC1(C(O)=O)C(=O)C2

InChI

1S/C10H14O3/c1-9(2)6-3-4-10(9,8(12)13)7(11)5-6/h6H,3-5H2,1-2H3,(H,12,13)/t6-,10+/m1/s1

InChI key

WDODWBQJVMBHCO-LDWIPMOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1S)-(+)-Ketopinic acid is a chiral ketone.

Application

(1S)-(+)-Ketopinic acid may be used to prepare:
  • Chiral phosphine-oxazoline ligands, which can used in asymmetric palladium-catalyzed Heck reaction of aryl or alkenyl triflates with cyclic alkenes.
  • A chiral imino-phosphine ligand, which can be used in palladium-catalyzed asymmetric Diels–Alder reactions to prepare six-membered carbocycles.
  • A chiral iminopyridine ligand, which can be used in copper-catalyzed Henry (nitro aldol) reaction between nitromethane and o-anisol to form the corresponding β-hydroxynitroalkane.
Used to prepare a new chiral oxazolidone auxiliary and a ketopinic derivatized polymer used for the deracemization of amines. Starting material for the synthesis of homochiral 2,10-camphanediols. Employed in the formation of a chiral Schiff base precursor to diethyl (S)-α-amino-α-alkyl phosphonates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yan, T.-H. et al.
Tetrahedron Letters, 32, 4959-4959 (1991)
Calmes, M. et al.
Tetrahedron Asymmetry, 5, 817-817 (1994)
Palladium-catalyzed asymmetric Diels-Alder reactions with novel chiral imino-phosphine ligands.
Hiroi K and Watanabe K.
Tetrahedron Asymmetry, 12(22), 3067-3071 (2001)
Chiral P, N-ligands based on ketopinic acid in the asymmetric Heck reaction.
Gilbertson SR and Fu Z.
Organic Letters, 3(2), 161-164 (2001)
M. Ferrari et al.
Synthetic Communications, 22, 107-107 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service