Skip to Content
Merck
All Photos(1)

Key Documents

41413

Sigma-Aldrich

2,6-Dimethylphenyl isocyanide

≥98.0% (GC)

Synonym(s):

vic.-m-Xylyl isocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NC
CAS Number:
Molecular Weight:
131.17
Beilstein:
3541909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

powder

mp

72-75 °C

storage temp.

2-8°C

SMILES string

Cc1cccc(C)c1[N+]#[C-]

InChI

1S/C9H9N/c1-7-5-4-6-8(2)9(7)10-3/h4-6H,1-2H3

InChI key

DNJLFZHMJDSJFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dimethylphenyl isocyanide may be employed for the synthesis of homoleptic metallaamidine complex, Mo(η2-Me2NCN-2,6-C6H3)4, via reaction with Mo(NMe2)4.

Other Notes

Review

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon-nitrogen bond formation in the reaction between tetrakis(dimethylamido)molybdenum(IV) and 2,6-dimethylphenyl isocyanide. Preparation and characterization of the first homoleptic metallaamidine complex: Mo(.eta.2-Me2NC:NC6H3Me3-2,6)4.
M H Chisholm et al.
Journal of the American Chemical Society, 108(24), 7860-7861 (1986-11-01)
Reactions of (Me3ECH2) 3ZrSi (SiMe3) 3 (E= C, Si) with 2, 6-Dimethylphenyl Isocyanide. Preferential Isocyanide Insertion into Zr-Silyl Bonds.
Wu Z, et al.
Organometallics, 17(22), 4853-4860 (1998)
Surface Enrichment of Ag Atoms in Au/Ag Alloy Nanoparticles Revealed by Surface-Enhanced Raman Scattering of 2, 6-Dimethylphenyl Isocyanide.
Kim K, et al.
The Journal of Physical Chemistry C, 114(8), 3448-3453 (2010)
M A Wood et al.
The Biochemical journal, 247(3), 675-678 (1987-11-01)
A series of aromatic isonitriles were synthesized and their binding to sheep haemoglobin and horse heart myoglobin was investigated. The disubstituted ligands 2,6-dimethylphenylisonitrile and 2,6-diethylphenylisonitrile were found to bind to horse-heart myoglobin with affinities ranging from 500 to 5000 times
M.P. Periassamy et al.
Organic preparations and procedures international, 11, 295-295 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service