Skip to Content
Merck
All Photos(2)

Key Documents

412228

Sigma-Aldrich

1-Pentadecanol

99%

Synonym(s):

Pentadecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14OH
CAS Number:
Molecular Weight:
228.41
Beilstein:
1700695
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

269-271 °C

mp

41-44 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCO

InChI

1S/C15H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h16H,2-15H2,1H3

InChI key

REIUXOLGHVXAEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pentadecanol is a linear alcohol. Vapor-phase composition of 1-pentadecanol in supercritical carbon dioxide at 323.15K has been evaluated by a flow-type apparatus. Molar heat capacities of 1-pentadecanol has been evaluated between 300K and 370K.

Application

1-Pentadecanol may be employed as starting reagent for the asymmetric synthesis of jaspine B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat Capacities and Derived Thermodynamic Functions of 1-Dodecanol and 1-Tridecanol between 10 K and 370 K and Heat Capacities of 1-Pentadecanol and 1-Heptadecanol between 300 K and 380 K and Correlations for the Heat Capacity and the Entropy of Liquid n-Alcohols.
van Miltenburg JC, et al.
Journal of Chemical and Engineering Data, 48(1), 36-43 (2003)
Vasiliki Gkarane et al.
Food research international (Ottawa, Ont.), 115, 54-64 (2019-01-03)
Animal production factors can affect the sensory quality of lamb meat. The study investigated the effect of diet composition and duration of consumption on the proximate analysis, volatile profile and sensory quality of lamb meat. Ninety-nine male Texel × Scottish
A novel stereoselective synthesis of pachastrissamine (jaspine B) starting from 1-pentadecanol.
Venkatesan K and Sreenivasan KV.
Tetrahedron Asymmetry, 19(2), 209-215 (2008)
Kalpana Shanmugarajah et al.
Scientific reports, 9(1), 11381-11381 (2019-08-08)
Diffusion barriers enable plant survival under fluctuating environmental conditions. They control internal water potential and protect against biotic or abiotic stress factors. How these protective molecules are deposited to the extracellular environment is poorly understood. We here examined the role
Jinrong Peng et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(5), 1700891-1700891 (2018-06-08)
The therapeutic outcome of photothermal therapy (PTT) remains impeded by the transparent depth of light. Combining PTT with immunotherapy provides strategies to solve this problem. Regulating metabolism-related enzymes is a promising strategy to stimulate immune response. Here, a nanosystem (NLG919/IR780

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service