Skip to Content
Merck
All Photos(1)

Key Documents

408964

Sigma-Aldrich

Cyclohexyl methacrylate

≥97%, contains ~60 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Methacrylic acid cyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOC6H11
CAS Number:
Molecular Weight:
168.23
Beilstein:
2045235
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

liquid

contains

~60 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.458 (lit.)

bp

68-70 °C/4 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OC1CCCCC1

InChI

1S/C10H16O2/c1-8(2)10(11)12-9-6-4-3-5-7-9/h9H,1,3-7H2,2H3

InChI key

OIWOHHBRDFKZNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexyl methacrylate(CHMA) is an acrylate monomer with a high-refractive index, commonly used in ultraviolet (UV) curing processes due to its ability to rapidly polymerize under UV light exposure. UV-curable CHMA-based coatings, adhesives, and inks offer fast curing, excellent hardness, hydrophobicity, and high durability. It has good optical clarity, resulting in optical lenses with high transparency and minimal distortion. It can also be used as a crosslinking agent in copolymer systems for dental composites and tissue engineering to enhance the mechanical and thermal properties of the resulting materials.

Application

Cyclohexyl methacrylate can be used:



  • As a monomer to prepare poly(cyclohexyl methacrylate) (PCHMA) thin films for high-performance luminescent solar concentrators(LSCs). These LSCs can be used to fabricate high-quantum yield photovoltaic solar cells.
  • As a comonomer to prepare photo-patternable quantum dot-acrylate resins for fabricating light-emitting diodes with high color transparency.
  • As a monomer to synthesizesuperhydrophobic silica nanoparticle surfaces for various applications such as anti-corrosionsurfaces, drag reduction, and energy conservation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High-Performance Luminescent Solar Concentrators Based on Poly (Cyclohexylmethacrylate)(PCHMA) Films
Francisco Jos'e Ostos, et al.
Polymers, 12, 2898-2898 (2020)
Azeotropic Binary Solvent System Containing Nonfluorinated Polymer-Grafted Silica Nanoparticles for the Fabrication of a Superhydrophobic Surface
Kai-Yen Chin, et al.
Langmuir, 39, 9841-9849 (2023)
Fabrication of light emitting diodes using photo-patternable quantum dot-acrylate resins
Kab Pil Yang, et al.
Journal of Materials Chemistry, 9, 17084-17088 (2021)
Florian Jakobs et al.
Materials (Basel, Switzerland), 13(8) (2020-04-29)
For most kinds of active polymer optical fibers, a homogeneous distribution of dye molecules over the entire fiber length and cross section is required. In this study, chemical bonding of dyes to poly(methyl methacrylate) (PMMA) by copolymerization is achieved within
Kai Kan et al.
Nanoscale research letters, 15(1), 51-51 (2020-03-04)
A characteristic effect of a nano-concave-convex structure of a zirconia nanoparticle assembly with an inherent porous structure and huge surface area enabled us to introduce systematic surface modification by thermal treatment to smooth surface and polymer impregnation to mask the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service