Skip to Content
Merck
All Photos(3)

Key Documents

402761

Sigma-Aldrich

Tin(II) oxalate

98%

Synonym(s):

Oxalic acid tin(II) salt, Stannous oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnC2O4
CAS Number:
Molecular Weight:
206.73
Beilstein:
3708588
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

core: tin
reagent type: catalyst

SMILES string

O=C1O[SnH2]OC1=O

InChI

1S/C2H2O4.Sn/c3-1(4)2(5)6;/h(H,3,4)(H,5,6);/q;+2/p-2

InChI key

OQBLGYCUQGDOOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1 - STOT RE 2

Target Organs

Cardio-vascular system, Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jung Sang Cho et al.
Scientific reports, 6, 23915-23915 (2016-04-02)
A commercially applicable and simple process for the preparation of aggregation-free metal oxide hollow nanospheres is developed by applying nanoscale Kirkendall diffusion to a large-scale spray drying process. The precursor powders prepared by spray drying are transformed into homogeneous metal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service