401803
Tetrabutylammonium acetate solution
1.0 M in H2O
Synonym(s):
N,N,N-tributyl-1-butanaminium acetate, Tetra-n-butylammonium acetate
About This Item
Recommended Products
form
liquid
Quality Level
concentration
1.0 M in H2O
refractive index
n20/D 1.389
bp
100 °C
density
0.99 g/mL at 25 °C
SMILES string
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
InChI
1S/C16H36N.C2H4O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2(3)4/h5-16H2,1-4H3;1H3,(H,3,4)/q+1;/p-1
InChI key
MCZDHTKJGDCTAE-UHFFFAOYSA-M
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service