Skip to Content
Merck
All Photos(1)

Key Documents

394882

Sigma-Aldrich

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide

>97%

Synonym(s):

MTBSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
241.33
Beilstein:
3606546
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade

Quality Level

Assay

>97%

form

liquid

refractive index

n20/D 1.402 (lit.)

bp

172-175 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

functional group

amine
fluoro

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C

InChI

1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3

InChI key

QRKUHYFDBWGLHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide is a highly reactive silylating reagent commonly used for silyation of carboxylic acids, alcohols, amines, and thiols. It is also used as a protection, deprotection reagent.

Application

N-tert-butyldimethylsilyl-N-methyltrifluoroacetamide (MTBSTFA) can be used:
  • As a derivatizing agent in the GC-MS analysis of hydroxylated fluorenes and in the GC analysis of amino acids.
  • As an efficient silylating agent for different alcohols, thiols, phenols, carboxylic acids, amines, and amides.
  • To functionalize multi-walled carbon nanotubes (MWCNTs) with polar groups.
MTBSTFA is reportedly much less moisture-sensitive than other silylating reagents used for GLC derivatization.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluating MTBSTFA derivatization reagents for measuring naphthenic acids by gas chromatography-mass spectrometry
Rozlyn YF,et al.
Analytical Methods, 2, 765-770 (2010)
Silylation of multi-walled carbon nanotubes
Aizawa M, et al.
Chemical Physics Letters, 368(1-2), 121-124 (2003)
High-throughput analysis of selected urinary hydroxy polycyclic aromatic hydrocarbons by an innovative automated solid-phase microextraction
Stefano D,et al.
Molecules (Basel), 23, 1869-1869 (2018)
Determination of β-blockers and β-agonists using gas chromatography and gas chromatography-mass spectrometry-a comparative study of the derivatization step
Caban M, et al.
Journal of Chromatography A, 1218(44), 8110-8122 (2011)
Two stage derivatization with N-(tert.-butyldimethylsilyl)-N-methyl-trifluoroacetamide (MTBSTFA) and N-methyl-bis-(trifluoroacetamide)(MBTFA) for the gas-chromatographic analysis of OH-, SH-and NH-compounds
Schoene K, et al.
Fresenius Journal of Analytical Chemistry, 348(5-6), 364-370 (1994)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service