394297
Magnesium fluoride
technical grade
Synonym(s):
Sellaite
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
MgF2
CAS Number:
Molecular Weight:
62.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
powder
Recommended Products
grade
technical grade
form
powder
density
3.15 g/mL at 25 °C (lit.)
SMILES string
F[Mg]F
InChI
1S/2FH.Mg/h2*1H;/q;;+2/p-2
InChI key
ORUIBWPALBXDOA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jonathan Lellouche et al.
International journal of nanomedicine, 7, 1175-1188 (2012-03-16)
The ability of bacteria to colonize catheters is a major cause of infection. In the current study, catheters were surface-modified with MgF(2) nanoparticles (NPs) using a sonochemical synthesis protocol described previously. The one-step synthesis and coating procedure yielded a homogenous
Jonathan Lellouche et al.
Biomaterials, 30(30), 5969-5978 (2009-08-12)
The ability of bacteria to develop antibiotic resistance and colonize abiotic surfaces by forming biofilms is a major cause of medical implant-associated infections and results in prolonged hospitalization periods and patient mortality. This raises the urgent need to develop compounds
Martina Thomann et al.
Journal of biomedical materials research. Part A, 93(4), 1609-1619 (2010-01-15)
MgCa0.8 cylinders (2.5 x 25 mm(2)) were coated with a magnesium-fluoride layer and implanted in the marrow cavities of both tibiae of 10 New Zealand White rabbits. The implantation duration was 3 and 6 months. The implants were clinically well
J Preben Morth et al.
Nature, 450(7172), 1043-1049 (2007-12-14)
The Na+,K+-ATPase generates electrochemical gradients for sodium and potassium that are vital to animal cells, exchanging three sodium ions for two potassium ions across the plasma membrane during each cycle of ATP hydrolysis. Here we present the X-ray crystal structure
Jonathan Lellouche et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(5), 702-711 (2011-09-29)
Antibiotic resistance has prompted the search for new agents that can inhibit bacterial growth. We recently reported on the antimicrobial and antibiofilm activities of nanosized magnesium fluoride (MgF(2)) nanoparticles (NPs) synthesized in ionic liquid using microwave chemistry. In this article
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service