Skip to Content
Merck
All Photos(1)

Key Documents

391662

Sigma-Aldrich

(1S,2S,3S,5R)-(+)-Isopinocampheylamine

95%

Synonym(s):

(+)-Isopinocampheylamine, (1S,2S,3S,5R)-3-Pinanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N
CAS Number:
Molecular Weight:
153.26
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

optical activity

[α]22/D +44°, neat

refractive index

n20/D 1.481 (lit.)

bp

90 °C/18 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1[C@@H](N)C[C@H]2C[C@@H]1C2(C)C

InChI

1S/C10H19N/c1-6-8-4-7(5-9(6)11)10(8,2)3/h6-9H,4-5,11H2,1-3H3/t6-,7+,8-,9-/m0/s1

InChI key

VPTSZLVPZCTAHZ-KZVJFYERSA-N

Application

(1S,2S,3S,5R)-(+)-isopinocampheylamine is a primary bicyclic amine with potent M2 ion channel inhibitor ability similar to that of amantadine, making it a promising candidate for developing anti-influenza agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New strategy for high throughput screening of anti-influenza virus M2 ion channel inhibitors.
Li C, et al.
Current Pharmaceutical Design, 19(28), 5146-5155 (2013)
Discovery of highly potent agents against influenza A virus.
Zhao X, et al.
European Journal of Medicinal Chemistry, 46(1), 52-57 (2011)
Identification of hits as matrix-2 protein inhibitors through the focused screening of a small primary amine library.
Hu W, et al.
Journal of Medicinal Chemistry, 53(9), 3831-3834 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service