Skip to Content
Merck
All Photos(2)

Key Documents

383481

Sigma-Aldrich

3-Cyanochromone

97%

Synonym(s):

4-Oxo-4H-1-benzopyran-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5NO2
CAS Number:
Molecular Weight:
171.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

174-176 °C (lit.)

functional group

ketone
nitrile

SMILES string

O=C1C(=COc2ccccc12)C#N

InChI

1S/C10H5NO2/c11-5-7-6-13-9-4-2-1-3-8(9)10(7)12/h1-4,6H

InChI key

SFWNPLLGXKJESA-UHFFFAOYSA-N

General description

3-Cyanochromone (4-Oxo-4H-1-benzopyran-3-carbonitrile), a 3-substituted chromone, is a cyano substituted 1-benzopyran-4-one. It is an α,β-unsaturated nitrile and also an α,β-unsaturated ketone. Its molecule has electron deficiency at 3 sites i.e C (carbon) at second position, C of cyano and carbonyl group. 2-Amino-3-(alkyliminomethyl)chromones are obtained as major products during its reaction with aliphatic amines.

Application

3-Cyanochromone may be used in the syntheses of following:
  • 2-aminochromone-3-carboxamide
  • 3-amino-4H-chromeno[3,4-d]isoxazol-4-one
  • 3-(diaminomethylene)chroman-2,4-dione
  • functionalized novel spirobenzofuranones
  • 6H-bis-[1]-benzopyrano[2,3-b:3′,4′-e]pyridin-8(8H)ones and 3-(2′-hydroxybenzoyl)-5H-[1]benzopyrano[4,3-b]pyridine

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS OF 6 H-bis-[1]-BENZOPYRANO [2, 3-b: 3', 4'-e] PYRIDIN-8 (8 H) ONES AND 3-(2'-HYDROXYBENZOYL)-5 H-[1] BENZOPYRANO [4, 3-b] PYRIDINES AND THEIR DERIVATIVES.
Reddy KV, et al.
Organic preparations and procedures international, 28(3), 325-332 (1996)
Structural revision in the reactions of 3-cyanochromones with primary aromatic amines. Improved synthesis of 2-amino-3-(aryliminomethyl) chromones.
Sosnovskikh VY, et al.
Tetrahedron Letters, 50(47), 6515-6518 (2009)
Tryfon Zarganes-Tzitzikas et al.
The Journal of organic chemistry, 76(21), 9008-9014 (2011-10-14)
Another aspect concerning chromone chemistry leading to the one-pot synthesis of functionalized novel spirobenzofuranones has been described. The synthesis involves reaction of the zwitterionic intermediates formed by the 1:1 interaction between isocyanides and acetylenecarboxylates with 3-cyanochromones, whereupon through an unexpected
Chemistry of 4-oxo-4H-1-benzopyran-3-carbonitrile.
Ghosh CK and Karak SK.
Journal of Heterocyclic Chemistry, 42(6), 1035-1035 (2005)
A reinvestigation of the reactions of 3-substituted chromones with hydroxylamine. Unexpected synthesis of 3-amino-4H-chromeno [3,4-d] isoxazol-4-one and 3-(diaminomethylene) chroman-2,4-dione.
Sosnovskikh VY, et al.
Tetrahedron Letters, 49(48), 6856-6859 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service