Skip to Content
Merck
All Photos(2)

Documents

380989

Sigma-Aldrich

Sodium perrhenate

99.99% trace metals basis

Synonym(s):

Sodium Rhenate(VII), Sodium rhenium oxide, Sodiumoxido(trioxo)rhenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaReO4
CAS Number:
Molecular Weight:
273.19
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder or crystals

impurities

≤150.0 ppm Trace Metal Analysis

density

5.39 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-][Re](=O)(=O)=O

InChI

1S/Na.4O.Re/q+1;;;;-1;

InChI key

KMISVFTVBOPTAI-UHFFFAOYSA-N

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dien Li et al.
Journal of hazardous materials, 374, 177-185 (2019-04-19)
Technetium-99 (99Tc) is a major contaminant at nuclear power plants and several US Department of Energy sites. Its most common aqueous species, pertechnetate (TcO4-), is very mobile in the environment, and currently there are no effective technologies for its sequestration.
J Tyler French et al.
Journal of vascular and interventional radiology : JVIR, 21(8), 1271-1279 (2010-05-19)
Minimally invasive interventional cancer therapy with drug-carrying lipid nanoparticles (ie, liposomes) via convection-enhanced delivery by an infusion pump can increase intratumoral drug concentration and retention while facilitating broad distribution throughout solid tumors. The authors investigated the utility of liposome-carrying beta-emitting
Michael J Booth et al.
Science (New York, N.Y.), 336(6083), 934-937 (2012-04-28)
5-Methylcytosine can be converted to 5-hydroxymethylcytosine (5hmC) in mammalian DNA by the ten-eleven translocation (TET) enzymes. We introduce oxidative bisulfite sequencing (oxBS-Seq), the first method for quantitative mapping of 5hmC in genomic DNA at single-nucleotide resolution. Selective chemical oxidation of
Albert Schröckeneder et al.
Inorganic chemistry, 48(24), 11608-11614 (2009-11-27)
Rhenium(V) oxo complexes of the type [ReOX(L)(2)] (1-7; X = Cl, Br) containing beta-ketiminate ligands (L = CH(3)C(O)CH(2)C(NAr)CH(3): Ar = Ph (APOH), 2-MePh (MPOH), 2,6-Me(2)Ph (DPOH), 2,6-(i)Pr(2)Ph (DiPOH)) have been prepared by reaction of [ReOX(3)(OPPh(3))(SMe(2))] (X = Cl, Br) with
Mark J Bailey et al.
Advanced healthcare materials, 1(4), 437-442 (2012-11-28)
Configuring RE(2) O(3) nanocrystals with pseudo-2D morphologies confers substantive gains in relaxivities over equivalent spherical counterparts. The most promising arises from Gd(2) O(3) whose ionic transverse and longitudinal relaxivities were as high as r(1) = 12.7 mM(-1) s(-1) and r(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service