Skip to Content
Merck
All Photos(2)

Key Documents

379379

Sigma-Aldrich

2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30O5
CAS Number:
Molecular Weight:
410.50
Beilstein:
4210983
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

SMILES string

CC(C)(C)c1cc(C(O)=O)c2Oc3c(cc(cc3C(C)(C)c2c1)C(C)(C)C)C(O)=O

InChI

1S/C25H30O5/c1-23(2,3)13-9-15(21(26)27)19-17(11-13)25(7,8)18-12-14(24(4,5)6)10-16(22(28)29)20(18)30-19/h9-12H,1-8H3,(H,26,27)(H,28,29)

InChI key

VJSNHUACDSJJCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid is a carboxylic acid building block.

Application

2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid may be used in the synthesis of 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbon­yl)-9H-xanthene methanol trisolvate monohydrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 7-Di-tert-butyl-9, 9-dimethyl-4, 5-bis (4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
Vysotsky MO and Schollmeyer D.
Acta Crystallographica Section E, Structure Reports Online, 63(1), o169-o171 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service