378119
Boron carbide
powder, <10 μm, 98%
Synonym(s):
Carbon tetraboride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CB4
CAS Number:
Molecular Weight:
55.25
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
98%
form
powder
particle size
<10 μm
density
2.51 g/mL at 25 °C (lit.)
SMILES string
B1=BB=C=B1
InChI
1S/CB4/c1-2-4-5-3-1
InChI key
NOJMLSPGQSYAIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Nanotwins soften boron-rich boron carbide (B13C2): This study examines the mechanical properties of nanotwinned boron-rich boron carbide, highlighting its potential applications in environments where materials need to combine toughness with hardness (Q An, WA Goddard, 2017).
- Structure and mechanical properties of boron-rich boron carbides: Analyzes the structure and mechanical properties of boron carbide, focusing on its exceptional hardness and potential applications in protective gear and abrasive materials (C Cheng et al., 2017).
- High density boron carbide ceramics: Discusses methods to achieve high-density boron carbide ceramics through hot pressing, relevant for applications requiring high material integrity (SN Perevislov et al., 2018).
- A review of tribological properties for boron carbide ceramics: Reviews the tribological properties of boron carbide, focusing on its wear resistance, an important property for drug discovery applications where material durability is crucial (W Zhang, 2021).
- Local atomic arrangements and band structure of boron carbide: Provides a detailed examination of the atomic structure and electronic properties of boron carbide, shedding light on its unique chemical properties useful for various chemical synthesis and materials science applications (K Rasim et al., 2018).
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yuqi Guo et al.
Nanoscale, 13(3), 1652-1662 (2021-01-12)
Liquid phase exfoliation (LPE) is a method that can be used to produce bulk quantities of two-dimensional (2D) nanosheets from layered van der Waals (vdW) materials. In recent years, LPE has been applied to several non-vdW materials with anisotropic bonding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service