Skip to Content
Merck
All Photos(2)

Key Documents

375195

Sigma-Aldrich

Trimethyltin chloride solution

1.0 M in THF

Synonym(s):

Chlorotrimethylstannane, Chlorotrimethyltin, Trimethylchlorostannane, Trimethylchlorotin, Trimethylstannyl chloride, Trimethyltin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnCl
CAS Number:
Molecular Weight:
199.27
Beilstein:
3535111
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in THF

density

0.988 g/mL at 25 °C

SMILES string

C[Sn](C)(C)Cl

InChI

1S/3CH3.ClH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

KWTSZCJMWHGPOS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erwin van Vliet et al.
Neurotoxicology, 28(6), 1136-1146 (2007-08-19)
Neurotoxicity aims to understand how xenobiotics interfere with the function of the nervous system and to unravel their mechanisms of action. Neuronal activity is the primary functional output of the nervous system and deviations from its resting level may indicate
Jiali Cai et al.
Chemical research in toxicology, 22(9), 1582-1587 (2009-08-07)
In evaluating the cytotoxic effects and the mechanisms of the apoptotic and necrotic actions of trimethyltin chloride (TMT) on human hepatoma G2 (HepG2) cells, the present study focused on the involvement of antiproliferation, DNA damage, cell death, apoptosis-related proteins, and
Guangshu Zhai et al.
Chemosphere, 72(3), 389-399 (2008-04-25)
The photodegradation of methyltins, as environmental pollutants, has scarcely been studied so far because of the shortage of rapid and sensitive speciation methods, even though they have very simple structures. The photodegradation of monomethyltin trichloride (MMT), dimethyltin dichloride (DMT) and
Mengmeng Wang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 22(5), 1136-1142 (2008-04-16)
In this study we intended to evaluate the cytotoxic and genotoxic effects of trimethyltin chloride (TMT), one of the most widely used organometallic compounds, on liver cells with the rat liver epithelial cell line IAR20. The results showed that TMT
Nguyen Quynh Huong et al.
Biological & pharmaceutical bulletin, 34(12), 1856-1863 (2011-12-02)
The organotin trimethyltin (TMT) is well known to cause neuronal degeneration in the hippocampal dentate gyrus of mice. The first purpose of the present study was to examine whether the cyclooxygenase (COX) inhibitor indomethacin could ameliorate neuronal degeneration in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service