Skip to Content
Merck
All Photos(3)

Key Documents

375160

Sigma-Aldrich

(S)-(+)-Ibuprofen

ReagentPlus®, 99%

Synonym(s):

S-(+)-IBU, (S)-(+)-2-(4-Isobutylphenyl)propionic acid, (S)-(+)-4-Isobutyl-α-methylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
206.28
Beilstein:
3590022
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

optical activity

[α]20/D +59°, c = 2 in ethanol

mp

49-53 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)Cc1ccc(cc1)[C@H](C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/t10-/m0/s1

InChI key

HEFNNWSXXWATRW-JTQLQIEISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(+)-Ibuprofen is the enantiomer associated with the anti-inflammatory action of ibuprofen, which is widely used as a nonsteroidal anti-inflammatory drug in racemic form.

Application

(S)-(+)-Ibuprofen may be used as a starting material to synthesize hydrogelators with the potential ability to release it via enzyme-mediated hydrolysis.
Enantiomeric form of the anti-inflammatory agent ibuprofen used in pharmacokinetic studies.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Jamali et al.
Journal of pharmaceutical sciences, 81(3), 221-225 (1992-03-01)
The influences of absorption rate and dosage size on the pharmacokinetics of ibuprofen (IB) enantiomers were studied in six healthy subjects. Rapidly absorbed solutions (50, 100, 200, 400, 600, and 1200 mg) and regular 600-mg tablets of racemic IB were
Enantioselective determination of ibuprofen in plasma by high-performance liquid chromatography-electrospray mass spectrometry.
Bonato PS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 796(2), 413-420 (2003)
(S)-(+)-Ibuprofen-based hydrogelators: an approach toward anti-inflammatory drug delivery.
Bhuniya S, et al.
Tetrahedron Letters, 47(40), 7153-7156 (2006)
Elizabeth R Southey et al.
Current medical research and opinion, 25(9), 2207-2222 (2009-07-18)
The main aim of this review was to compare the tolerability and safety between ibuprofen and paracetamol when used as anti-pyretic and analgesic agents in children up to 18 years of age. MEDLINE (1950 to November 2008), EMBASE (1980 to
Jennifer Manley et al.
The Annals of pharmacotherapy, 41(7), 1227-1232 (2007-05-24)
To evaluate the literature examining prophylactic use of acetaminophen and ibuprofen for prevention of adverse reactions associated with childhood immunization. Articles were identified via MEDLINE/PubMed/EMBASE (1966-March 2007) using the following key terms: vaccination, immunization, diphtheria-tetanus toxoids-whole pertussis (DTwP), diphtheria tetanus-toxoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service