Skip to Content
Merck
All Photos(2)

Key Documents

375004

Sigma-Aldrich

2,4-Thiazolidinedione

technical grade, 90%

Synonym(s):

2,4(3H,5H)-Thiazoledione, 2,4-Dioxo-1,3-thiazolidine, 2,4-Dioxothiazolidine, 2-Hydroxy-4,5-dihydro-1,3-thiazol-4-one, Thiazole-2,4-dione, Thiazolidin-2,4-dione, Thiazolidinedione (6CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3NO2S
CAS Number:
Molecular Weight:
117.13
Beilstein:
110700
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

mp

125-127 °C (lit.)

SMILES string

O=C1CSC(=O)N1

InChI

1S/C3H3NO2S/c5-2-1-7-3(6)4-2/h1H2,(H,4,5,6)

InChI key

ZOBPZXTWZATXDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Thiazolidinedione is a well-known peroxisome proliferator-activated receptors (PPAR) activator. Its effect on aging process using kidneys from Fischer 344 rats has been reported. A library of conjugates of 2,4-thiazolidinedione has been synthesized by Knoevenagel condensation followed by reduction using hydrogen gas and Pd/C catalyst. Knoevenagel condensation of various aromatic aldehydes with 2,4-thiazolidinedione in polyethylene glycol-300 has been reported.

Application

2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K M Schoenberg et al.
Journal of dairy science, 94(11), 5523-5532 (2011-10-29)
Administration of peroxisome proliferator-activated receptor gamma (PPARγ) ligands, thiazolidinediones (TZD), to prepartum dairy cattle has been shown to improve dry matter intake and decrease circulating nonesterified fatty acids (NEFA) around the time of calving. The objective of this work was
Jaelim Cho et al.
Clinical pharmacology and therapeutics, 107(3), 580-587 (2019-09-26)
Acute pancreatitis (AP) often progresses to recurrent acute pancreatitis (RAP) and chronic pancreatitis (CP). We investigated the relationship between the use of insulin after AP and progression from AP to RAP or CP, as well as the effect of diabetes
Shana Jaaf et al.
Veterinary sciences, 6(2) (2019-05-22)
Background: In a prior experiment, treatment of goats with the putative PPARγ agonist 2,4-thiazolidinedione (2,4-TZD) did not affect milk fat or expression of milk-fat related genes. The lack of response was possibly due to deficiency of vitamin A and/or a
Oscar Lamas Longarela et al.
PloS one, 8(3), e58340-e58340 (2013-03-19)
The hepatitis delta virus (HDV) is a small, defective RNA virus that requires the presence of the hepatitis B virus (HBV) for its life cycle. Worldwide more than 15 million people are co-infected with HBV and HDV. Although much effort
Sabrina Heng et al.
Bioorganic & medicinal chemistry, 19(24), 7453-7463 (2011-11-15)
We present a new class of inhibitors of pancreatic cholesterol esterase (CEase) based on 'priviledged' 5-benzylidenerhodanine and 5-benzylidene-2,4-thiazolidinedione structural scaffolds. The lead structures (5-benzylidenerhodanine 4a and 5-benzylidene-2,4-thiazolidinedione 4b) were identified in an in-house screening and these inhibited CEase with some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service