Skip to Content
Merck
All Photos(2)

Key Documents

370932

Sigma-Aldrich

Hydroxypropyl acrylate, mixture of isomers

contains 200 ppm hydroquinone monomethyl ether as inhibitor, 95%

Synonym(s):

Propylene glycol acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2C3H6OH
CAS Number:
Molecular Weight:
130.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

4.5 (vs air)

Quality Level

Assay

95%

form

liquid

contains

200 ppm hydroquinone monomethyl ether as inhibitor

refractive index

n20/D 1.445 (lit.)

bp

77 °C/5 mmHg (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

CC(O)COC(=O)C=C.CC(CO)OC(=O)C=C

InChI

1S/2C6H10O3/c1-3-6(8)9-4-5(2)7;1-3-6(8)9-5(2)4-7/h2*3,5,7H,1,4H2,2H3

InChI key

LCAYFFNMFIVNDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydroxypropyl acrylates may be used for the synthesis of a number of acrylates. The presence of both vinyl and hydroxyl allows copolymerization of the acrylate with vinyl monomers as well as with di- or polyisocyanates, anhydrides, or epoxides.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contact dermatitis from the irritancy (immediate and delayed) and allergenicity of hydroxypropyl acrylate.
C R Lovell et al.
Contact dermatitis, 12(2), 117-118 (1985-02-01)
Reaction kinetics of the catalytic esterification of acrylic acid with propylene glycol.
Alt?okka MR and Odes E
Applied Catalysis A: General, 362(1), 115-120 (2009)
Rahul P Dixit et al.
Pharmaceutical development and technology, 12(5), 495-504 (2007-10-27)
In the present study, Simvastatin was incorporated in emulsion of soybean oil and propylene glycol monocaprylate as oily phase and Tween 80 and Cremophor EL as surfactants and also their mixtures. Dry adsorbed emulsions were prepared by using colloidal silicon
[Determination of propylene glycol monoacrylate and monomethacrylate propylene oxide in the air].
E Sh Gronsberg
Gigiena truda i professional'nye zabolevaniia, (4)(4), 48-49 (1981-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service