Skip to Content
Merck
All Photos(1)

Documents

367583

Sigma-Aldrich

4-Bromo-2-fluorobiphenyl

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(Br)F
CAS Number:
Molecular Weight:
251.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

39-41 °C (lit.)

SMILES string

Fc1cc(Br)ccc1-c2ccccc2

InChI

1S/C12H8BrF/c13-10-6-7-11(12(14)8-10)9-4-2-1-3-5-9/h1-8H

InChI key

HTRNHWBOBYFTQF-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 4-Bromo-2-fluorobiphenyl by Suzuki Coupling Reaction with Pd (PPh_3) _4 Catalysis.
Gao F-q, et al.
Fine Chemicals / ????, 11, 023-023 (2012)
Comparison of the properties of liquid crystals derived from certain lateral halogeno-substituted azomethines.
Brown JW, et al.
Mol. Cryst. Liq. Cryst., 159(1), 255-266 (1988)
The Research on the Synthetic Craft of 4-Bromo-2-Fluoro Biphenyl.
Wang C-l, et al.
Journal of Jiangxi Normal University (Natural Science Edition), 2, 010-010 (2007)
Ou-Ya Zhao et al.
Chemosphere, 174, 774-780 (2017-02-16)
The present study used strain ZH-H2 (Fusarium sp.) isolated by our group as the PAH-degrading strain and 5-6-rings PAHs as degradation objects. The soil incubation experiment was carried out to investigate the starch-enhanced degradation effects of HMW PAHs by Fusarium
Xiaoxue Zhang et al.
Environmental science and pollution research international, 27(34), 42969-42978 (2020-07-30)
The capacity of Fusarium sp. strain ZH-H2 to secret lignin peroxidase (LiP), laccase (Lac), and manganese peroxidase (MnP) and degrade high molecular weight polycyclic aromatic hydrocarbons (HMW-PAHs) was studied. When the fungus was grown in control mineral salt medium for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service