Skip to Content
Merck
All Photos(2)

Documents

363464

Sigma-Aldrich

1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18Cl2N2 · HCl
CAS Number:
Molecular Weight:
309.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

198-203 °C (lit.)

SMILES string

Cl[H].ClCCCN1CCN(CC1)c2cccc(Cl)c2

InChI

1S/C13H18Cl2N2.ClH/c14-5-2-6-16-7-9-17(10-8-16)13-4-1-3-12(15)11-13;/h1,3-4,11H,2,5-10H2;1H

InChI key

JVLRNANYLVRULL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride is present as impurity in nefazodone hydrochloride (pharmaceutical formulation). Synthesis of 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride is reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1-(3-Chlorophenyl)-4-(3-Chloropropyl)-Piperazine Hydrochloride.
<BIG>Mai-Chun S, et al. </BIG>
Fine Chemicals / ????, 35(5) (2005)
D Sreenivas Rao et al.
Journal of pharmaceutical and biomedical analysis, 26(4), 629-636 (2001-08-23)
A simple, selective and reproducible reversed-phase liquid chromatography (LC) method has been developed for the quantitative determination of nefazodone hydrochloride (I) in the presence of its related impurities, namely 5-ethyl-4-(2-phenoxyethyl)-2H-1,2,4-triazol-3-(4H)one (II), 1-(3-chlorophenyl)-4-(3-chloropropyl) piperazine hydrochloride (III) and 1,1(1)-trimethylene-bis[4-(3-chlorophenyl) piperazine] hydrochloride (IV).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service