Skip to Content
Merck
All Photos(1)

Documents

347795

Sigma-Aldrich

Sodium dichloroacetate

98%

Synonym(s):

Dichloroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCO2Na
CAS Number:
Molecular Weight:
150.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

impurities

<2% ethyl alcohol

mp

198 °C (dec.) (lit.)

SMILES string

[Na+].[O-]C(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2.Na/c3-1(4)2(5)6;/h1H,(H,5,6);/q;+1/p-1

InChI key

LUPNKHXLFSSUGS-UHFFFAOYSA-M

Gene Information

General description

Sodium dichloroacetate is a mitochondrial pyruvate dehydrogenase kinase (PDK) inhibitor that exhibits potent anti-leukemic activity.

Application

Sodium dichloroacetate can be used as an organic salt precursor for synthesizing microporous carbon that is useful in developing carbon-based adsorbents.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Ammendola et al.
Scientific reports, 10(1), 6521-6521 (2020-04-18)
The growing interest of oncologists in natural compounds such as polyphenols and flavonoids is encouraging the development of innovative and efficient carriers for the delivery of those drugs. This study examines carboxymethyl chitosan-based microcapsules created by spray drying as a
Preparing microporous carbon from solid organic salt precursors using in situ templating and a fixed-bed reactor.
Atkinson JD & Rood MJ.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 160, 174-181 (2012)
Mito-DCA: a mitochondria targeted molecular scaffold for efficacious delivery of metabolic modulator dichloroacetate.
Pathak RK, et al.
ACS Chemical Biology, 9(5), 1178-1187 (2014)
Yongchong Yang et al.
European journal of medicinal chemistry, 45(9), 4300-4306 (2010-07-29)
A current study shows that sodium dichloroacetate (DCA) can induce cancer cell apoptosis and inhibit tumor growth, but its cytotoxic activity is low (IC(50) > 1000 microM for A549). In this paper, a variety of DCA derivatives were synthesized, and
Peng Wang et al.
Shock (Augusta, Ga.), 49(6), 704-711 (2017-08-29)
No pharmacological interventions are currently available to provide neuroprotection for patients suffering from cardiac arrest. Dichloroacetate (DCA) is a pyruvate dehydrogenase kinase inhibitor, which activates pyruvate dehydrogenase (PDH), and increases cell adenosine triphosphate (ATP) production by promoting influx of pyruvate

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service