Skip to Content
Merck
All Photos(2)

Key Documents

342297

Sigma-Aldrich

2,7-Dibromofluorene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Br2
CAS Number:
Molecular Weight:
324.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Quality Level

Assay

97%

mp

164-166 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc-2c(Cc3cc(Br)ccc-23)c1

InChI

1S/C13H8Br2/c14-10-1-3-12-8(6-10)5-9-7-11(15)2-4-13(9)12/h1-4,6-7H,5H2

InChI key

AVXFJPFSWLMKSG-UHFFFAOYSA-N

General description

2,7-Dibromofluorene is a halogenated polycyclic aromatic compound and its vapour pressure has been measured using the Knudsen effusion method.

Application

2,7-Dibromofluorene was used as a template for the N-carbazole capped oligofluorenes which show potential as hole-transporting materials for organic light emitting devices (OLEDs). It may be used in the synthesis of conjugated polymer, poly[9,9′-bis(6′′-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), used in label-free DNA microarrays. It was used in the preparartion of blue photoluminescent unsymmetrically substituted polyfluorene. It was also used in the preparation of 2,7-dibromofluorene monomers containing benzyl ether dendrons (generations 1, 2 and 3) in the 9,9′-position of the fluorene ring, such as:
  • 9,9-bis[(3,5-bis(benzyloxy)benzyloxy)methyl]-2,7-dibromofluorene
  • 9,9-bis[(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy)-methyl] 2,7-dibromo-fluorene
  • 9,9-bis[(3,5-bis(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy) benzyloxy)-methyl]-2,7-dibromofluorene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Z Tang et al.
Chemical communications (Cambridge, England), (23)(23), 2426-2427 (2002-09-25)
The first unsymmetrically substituted polyfluorene bearing a bulky poly(benzyl ether) dendron and less bulky 3,6-dioxaoctyl groups in the 9-position was designed and synthesized, which gives almost a pure bluish photoluminescence with negligible weak greenish excimer emission around 520 nm even
D Marsitzky et al.
Journal of the American Chemical Society, 123(29), 6965-6972 (2001-07-19)
The synthesis and characterization of complex dendritic, rigid rod poly-2,7-fluorene homopolymers and copolymers via a macromonomer approach is reported. Several 2,7-dibromofluorene monomers containing benzyl ether dendrons (generations 1, 2, and 3) in the 9,9'-position of the fluorene ring were prepared
Jinxia Fu et al.
Environmental toxicology and chemistry, 31(3), 486-493 (2011-12-06)
Knowledge of vapor pressure of organic pollutants is essential in predicting their fate and transport in the environment. In the present study, the vapor pressures of 12 halogenated polycyclic aromatic compounds (PACs), 9-chlorofluorene, 2,7-dichlorofluorene, 2-bromofluorene, 9-bromofluorene, 2,7-dibromofluorene, 2-bromoanthracene, 9-chlorophenanthrene, 9-bromophenanthrene
Tetrahedron Letters, 48, 89-89 (2007)
Bin Liu et al.
Nature protocols, 1(4), 1698-1702 (2007-05-10)
We describe the synthesis of poly[9,9'-bis(6''-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), a cationic, water-soluble conjugated polymer used in label-free DNA microarrays. This polymer was designed to have a maximum absorbance of close to 488 nm, which meets the excitation wavelength of most commercial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service