Skip to Content
Merck
All Photos(2)

Key Documents

337374

Sigma-Aldrich

Antimony(III) chloride

≥99.95% trace metals basis

Synonym(s):

Antimony trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl3
CAS Number:
Molecular Weight:
228.12
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38080204
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.95% trace metals basis
form:
crystalline

vapor density

7.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 49 °C)

Assay

≥99.95% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: antimony

impurities

≤500.0 ppm Trace Metal Analysis

mp

73.4 °C (lit.)

SMILES string

Cl[Sb](Cl)Cl

InChI

1S/3ClH.Sb/h3*1H;/q;;;+3/p-3

InChI key

FAPDDOBMIUGHIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

  • Obtaining of nanosized compound Sb2S3: This study focuses on the formation of Sb2S3 nanorods using antimony(III) chloride, relevant for material science applications in semiconductors and sensors (Guseynov et al., 2017).
  • Highly Efficient Light‐Emitting Diodes Based on an Organic Antimony (III) Halide Hybrid: This study presents applications in optoelectronics, specifically in the development of efficient light-emitting diodes, of interest to material science chemists (Li et al., 2022).
  • Antimony (III) halide compounds of thioureas: Structures and biological activity: This research explores the biological activity of antimony(III) halide complexes, which could be of interest to drug discovery chemists looking for novel therapeutic agents (Hadjikakou et al., 2014).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tahani Alqahtani et al.
Scientific reports, 11(1), 1887-1887 (2021-01-23)
We report a simple, economical and low temperature route for phase-pure synthesis of two distinct phases of Cu-Sb-S, chalcostibite (CuSbS2) and tetrahedrite (Cu12Sb4S13) nanostructures. Both compounds were prepared by the decomposition of a mixture of bis(O-ethylxanthato)copper(II) and tris(O-ethylxanthato)antimony(III), without the use
James M Hogg et al.
Dalton transactions (Cambridge, England : 2003), 46(35), 11561-11574 (2017-08-03)
Coordination complexes of Lewis acidic metal chlorides AlCl
N Schaumlöffel et al.
Mutagenesis, 13(3), 281-286 (1998-06-27)
Data on the mechanism of antimony genotoxicity is scarce. Arsenic and antimony are proposed to share some toxicological features. Thus comparative and combined experiments with As(III) and Sb(III) were performed to gain a deeper knowledge of the mechanism of antimony
Why not Nichols chemosurgery?
Gary A Dyer
Clinics in dermatology, 24(5), 458-460 (2006-09-13)
S A Hussain et al.
Human & experimental toxicology, 17(3), 140-143 (1998-05-20)
1. The toxic gas hypothesis proposes exposure to stibine (antimony trihydride) generated from microbial contamination of cot mattress materials as a possible cause of unexplained death in infancy (SIDS) as a consequence of cholinesterase inhibition. We have measured the direct

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service