Skip to Content
Merck
All Photos(2)

Documents

333492

Sigma-Aldrich

Titanium(IV) sulfide

powder, -200 mesh, 99.9%

Synonym(s):

Titanium disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiS2
CAS Number:
Molecular Weight:
112.00
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9%

form

powder

reaction suitability

core: titanium
reagent type: catalyst

particle size

-200 mesh

SMILES string

S=[Ti]=S

InChI

1S/2S.Ti

InChI key

CFJRPNFOLVDFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV)sulfide (TiS2) is a commercially available transition metal sulfide catalyst. TiS2 is classified as a transition metal dichalcogenide and used in rechargeable batteries as a cathode material.

Application

Titanium(IV)sulfide is used to analyze oxidation in various environmental conditions using advanced electron microscopy.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-heat. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experimental evaluation of the effect of aerosols of titanium mono- and disulfide on the body].
I T Brakhnova et al.
Gigiena truda i professional'nye zabolevaniia, (8)(8), 50-52 (1989-01-01)
Growth of noble metal nanoparticles on single-layer TiS 2 and TaS 2 nanosheets for hydrogen evolution reaction
Zeng Z, et al.
Energy & Environmental Science, 797-803, 7(2)-7(2) (2014)
An in situ and ex situ TEM study into the oxidation of titanium (IV) sulphide
Long E, et al.
NPJ 2D materials and applications, 1(1), 1-9 (2017)
Ying Wen Linda Lim et al.
Nanoscale, 2(12), 2751-2757 (2010-10-13)
Titania (TiO2) and sodium titanate nanostructures with controllable phases and morphologies were synthesized by a hydrothermal method with titanium disulfide (TiS2) as the starting material. Sodium titanate nanobelts could be synthesized under a relatively low alkaline concentration (1 mol L(-1)
Sujay Prabakar et al.
Chemical communications (Cambridge, England), 47(1), 439-441 (2010-09-21)
The synthesis of inorganic fullerene (IF) nanoparticles and IF hollow spheres of titanium disulfide by a simple colloidal route is reported. The injection temperature of the titanium precursor into the solvent mixture was found to be important in controlling the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service