Skip to Content
Merck
All Photos(1)

Key Documents

333115

Sigma-Aldrich

2-Isopropylmalic acid

98%

Synonym(s):

2-Hydroxy-2-isopropylsuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C[CH(CH3)2](OH)CO2H
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

144-146 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

CC(C)C(O)(CC(O)=O)C(O)=O

InChI

1S/C7H12O5/c1-4(2)7(12,6(10)11)3-5(8)9/h4,12H,3H2,1-2H3,(H,8,9)(H,10,11)

InChI key

BITYXLXUCSKTJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Isopropylmalic acid is an intermediate formed during leucine biosynthesis. Budding yeast Saccharomyces cerevisiae secrets 2-isopropylmalic acid and helps in reduction of aluminum toxicity by forming chelate with Al ions, thereby preventing them from entering the cells.

Application

2-Isopropylmalic acid was employed as an internal standard in diagnostic model for pancreatic cancer using GC/MS-based human serum metabolomics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlo M T Marobbio et al.
The Journal of biological chemistry, 283(42), 28445-28453 (2008-08-07)
In Saccharomyces cerevisiae, alpha-isopropylmalate (alpha-IPM), which is produced in mitochondria, must be exported to the cytosol where it is required for leucine biosynthesis. Recombinant and reconstituted mitochondrial oxalacetate carrier (Oac1p) efficiently transported alpha-IPM in addition to its known substrates oxalacetate
Taisuke Suzuki et al.
Biological trace element research, 120(1-3), 257-263 (2007-10-06)
The budding yeast Saccharomyces cerevisiae secretes 2-isopropylmalic acid (2-iPMA), an intermediate in leucine biosynthesis. Because 2-iPMA binds Al(III) in the culture medium, it is thought to reduce toxicity by Al(III). The effects of 2-iPMA and malic acid (MA) on Al
Shozo Tomonaga et al.
Animal science journal = Nihon chikusan Gakkaiho, 89(2), 448-455 (2017-11-21)
Exposure to high ambient temperature is detrimental to the poultry industry. To understand the influence from a metabolic perspective, we investigated the effects of exposure to high ambient temperature on plasma low-molecular-weight metabolite levels in chicks using gas chromatography/mass spectrometry-based
P Friden et al.
Molecular and cellular biology, 9(9), 4056-4060 (1989-09-01)
LEU3 of Saccharomyces cerevisiae encodes an 886-amino-acid polypeptide that activates transcription of at least five genes by binding to an upstream decanucleotide sequence. This activation is dependent on the inducer alpha-isopropylmalate, the synthesis of which is repressed by leucine. We
P S Nielsen et al.
Molecular & general genetics : MGG, 264(5), 613-622 (2001-02-24)
Uptake of branched-chain amino acids by Saccharomyces cerevisiae from media containing a preferred nitrogen source is mediated by the permeases encoded by BAP2, BAP3, and VAP1/TAT1. The transcriptional activity of the BAP2 promoter is affected by a number of genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service