All Photos(1)
About This Item
Linear Formula:
CH3CH(OH)CH2SH
CAS Number:
Molecular Weight:
92.16
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.486 (lit.)
bp
58-60 °C/17 mmHg (lit.)
density
1.048 g/mL at 25 °C (lit.)
SMILES string
CC(O)CS
InChI
1S/C3H8OS/c1-3(4)2-5/h3-5H,2H2,1H3
InChI key
FETFXNFGOYOOSP-UHFFFAOYSA-N
Related Categories
General description
1-Mercapto-2-propanol is a chain transferring agent that is the sixth series of hydrogen bonded 1,2-propanols which can be used to form polymeric chains with low molecular weight and short chain length.
Application
1-Mercapto-2-propanol can be used as a self-assembled monolayer on the gold surfaces with potential use in electrochemical applications. It may also be used as a chiral agent which can be used in the chemical analysis of enantiomers.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
145.4 °F - closed cup
Flash Point(C)
63 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Microwave spectrum, conformation and intramolecular hydrogen bonding of 1-mercapto-2-propanol
Marstokk KM, et al.
Acta Chemica Scandinavica, 44, 339-345 (1990)
Non-local spatial frequency response of photopolymer materials containing chain transfer agents: II. Experimental results.
Guo J, et al.
Journal of Optics, 13(9), 095602-095602 (2011)
Microwave Spectrum, Conformation and Intramolecular Hydrogen Bonding of 1-Mercapto-2-propanol.
Marstokk KM, et al.
Acta Chemica Scandinavica, 44(9), 339-345 (1990)
Structural evolution of self-assembled monolayer of 1-mercapto-2-propanol on Au (1 1 1) in a N2 flow: an electrochemical and STM study.
Rhee CK and Kim YN
Applied Surface Science, 228(1-4), 313-319 (2004)
Combinatorial Synthesis and Screening of Novel Odorants Such as Polyfunctional Thiols
Vermeulen C and Collin S
Combinatorial Chemistry & High Throughput Screening, 9, 583-590 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service