Skip to Content
Merck
All Photos(1)

Documents

32784

Sigma-Aldrich

4,4′-Ethylenedianiline

technical, ≥95% (NT)

Synonym(s):

α,α′-Bi-p-toluidine, 4,4′-Diaminobibenzyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2
CAS Number:
Molecular Weight:
212.29
Beilstein:
394999
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥95% (NT)

form

solid

mp

133-137 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc(CCc2ccc(N)cc2)cc1

InChI

1S/C14H16N2/c15-13-7-3-11(4-8-13)1-2-12-5-9-14(16)10-6-12/h3-10H,1-2,15-16H2

InChI key

UHNUHZHQLCGZDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Ethylenedianiline may be used as an internal standard for the determination of 4,4′-methylenedianiline in urine samples using gas chromatography coupled with mass spectrometry (GC-MS).
4,4′-Ethylenedianiline was used in preparation of molecularly imprinting sol-gel materials for detection of DDT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of 4,4?-methylenedianiline in human urine.
Peterson JC, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 564(1), 205-212 (1991)
Amy L Graham et al.
Analytical chemistry, 74(2), 458-467 (2002-01-29)
Molecularly imprinting sol-gel materials for DDT using both a noncovalent and a covalent approach was examined. A nonpolar porous sol-gel network was created through the use of the bridged polysilsesquioxane, bis-(trimethoxysilylethyl)benzene (BTEB), as the principal sol-gel component. Noncovalent molecular imprinting
Petros Giastas et al.
Acta crystallographica. Section B, Structural science, 59(Pt 2), 287-299 (2003-03-27)
The present investigation is part of an ongoing study on the influence of the long end-functonalized guest molecules DBA and BNZ in the crystal packing of beta-cyclodextrin (betaCD) dimeric complexes. The title compounds are 2:2 host:guest complexes showing limited host-guest
X Wu et al.
Se pu = Chinese journal of chromatography, 16(6), 536-538 (2002-04-10)
A method for simultaneous determination of trace 4,4'-diaminobiphenyl, 4-nitrophenol and phenol in waste water by high performance liquid chromatography with spectrophotometric detection is reported. These components were separated on a Phenomenex Spherex C18 column with V(acetonitrile): V(ether): V(50 mmol.L-1 acetate)
[Determination of the carcinogenic risk of diaminodibenzyl (DADB)].
M Balan et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 84(3), 521-525 (1980-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service