Skip to Content
Merck
All Photos(1)

Key Documents

327212

Sigma-Aldrich

Paraffin wax

mp 58-62 °C (ASTM D 87)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mp

58-62 °C (ASTM D 87)

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Paraffin wax is a low cost petroleum product that contains hydrocarbons with 20-40 carbon atoms. It is mainly used as lubricant, candles and electrical insulation.

Application

Paraffin wax can be used as a phase changing material in a wide range of applications which include solar based water heaters, microcapsules and thermal energy devices(TEDs).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paraffin Products, 14 (1983)
Performance investigation of thermal energy storage system with Phase Change Material (PCM) for solar water heating application.
Mahfuz MH, et al.
International Communications in Heat and Mass Transfer, 57(3), 132-139 (2014)
Development of thermo-regulating textiles using paraffin wax microcapsules.
Sanchez P, et al.
Thermochimica Acta, 498(1-2), 16-21 (2010)
Heat transfer enhancement of paraffin wax using graphite foam for thermal energy storage.
Zhong Y, et al
Solar Energy Materials and Solar Cells, 94(6), 1011-1014 (2010)
Compounding of ethylene-propylene polymers for electrical applications.
Brown M.
IEEE Electrical Insulation Magazine, 10(1), 16-22 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service