323659
1-Fluoropyridinium triflate
99%
Synonym(s):
1-Fluoropyridinium trifluoromethanesulfonate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H5F4NO3S
CAS Number:
Molecular Weight:
247.17
Beilstein:
4222821
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
mp
185-186 °C (lit.)
functional group
fluoro
triflate
SMILES string
F[n+]1ccccc1.[O-]S(=O)(=O)C(F)(F)F
InChI
1S/C5H5FN.CHF3O3S/c6-7-4-2-1-3-5-7;2-1(3,4)8(5,6)7/h1-5H;(H,5,6,7)/q+1;/p-1
InChI key
JFZMMCYRTJBQQI-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
General purpose electrophilic fluorinating reagent.
Reactant for:
- Pd-catalyzed arylation reactions
- Preparation of small molecule botulinum neurotoxin A endopeptidase inhibitors
- Fluorination agent
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aldrichimica Acta, 28, 36-36 (1995)
Development of GSKDevelopment of GSK's reagent guides ? embedding sustainability into reagent selections reagent guides ? embedding sustainability into reagent selection.
Adams JP, et al.
Green Chemistry, 15, 1542-1549 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service