Skip to Content
Merck
All Photos(1)

Key Documents

317292

Sigma-Aldrich

Triethylene glycol monomethyl ether

95%

Synonym(s):

Methyltriglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)3OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1700198
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

5.66 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

95%

form

liquid

impurities

~2% corresponding tetraethylene glycol ethers
~2% each of the corresponding diethylene glycol methyl ether

refractive index

n20/D 1.439 (lit.)

bp

122 °C/10 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOCCO

InChI

1S/C7H16O4/c1-9-4-5-11-7-6-10-3-2-8/h8H,2-7H2,1H3

InChI key

JLGLQAWTXXGVEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylene glycol monomethyl ether can be used as a reagent and solvent for applications such as:
  • modification of anthraquinone material for redox flow batteries
  • preparation of polymeric electrolyte for electrochemical devices,
  • formation of the binary system of polyethylene glycol for absorption of silica.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthraquinone with tailored structure for a nonaqueous metal-organic redox flow battery
Wang W, et al.
Chemical Communications (Cambridge, England), 48(53), 6669-6671 (2012)
Highly efficient SO2 absorption and its subsequent utilization by weak base/polyethylene glycol binary system
Yang Z, et al.
Environmental Science & Technology, 47(3), 1598-1605 (2013)
Siloxyaluminate polymers with high Li+ ion conductivity
Fujinami T, et al.
Chemistry of Materials, 9(10), 2236-2239 (1997)
Benjamin B Rich et al.
Soft matter, 14(38), 7782-7792 (2018-08-07)
Plants are protected from the elements by a complex hierarchical epicuticular wax layer which has inspired the creation of super-hydrophobic and self-cleaning surfaces. Although many studies have been conducted on different plant wax systems to determine the mechanisms of water
Bapurao Surnar et al.
Nanoscale, 7(42), 17964-17979 (2015-10-16)
Platinum drug delivery against the detoxification of cytoplasmic thiols is urgently required for achieving efficacy in breast cancer treatment that is over expressed by glutathione (GSH, thiol-oligopeptide). GSH-resistant polymer-cisplatin core-shell nanoparticles were custom designed based on biodegradable carboxylic functional polycaprolactone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service