Skip to Content
Merck
All Photos(1)

Documents

316555

Sigma-Aldrich

1,6-Anhydro-β-D-glucose

99%

Synonym(s):

Levoglucosan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
Beilstein:
80998
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder or crystals

optical activity

[α]18/D −66°, c = 1 in H2O

color

beige

mp

182-184 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

O[C@H]1[C@H](O)[C@H]2CO[C@H](O2)[C@@H]1O

InChI

1S/C6H10O5/c7-3-2-1-10-6(11-2)5(9)4(3)8/h2-9H,1H2/t2-,3-,4+,5-,6-/m1/s1

InChI key

TWNIBLMWSKIRAT-VFUOTHLCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,6-Anhydro-β-D-glucose (Levoglucosan) can be used as a precursor for the synthesis of:
  • 1,6-anhydro-2,4-dideoxy-β-D-glycerohexopyranos-3-ulose.
  • Quasi-linear polyglucose (PGlc) via cationic ring-opening polymerization.
  • Polynitrogenated analogs of glucopyranoses.
  • 3-deoxy-3-fluorogalactopyranose and acetylated 4-deoxy-4-fluorogalactopyranose.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitra Balla et al.
Environmental science and pollution research international, 25(13), 12191-12205 (2017-09-10)
Two classes of polar organic compounds, dicarboxylic acids (DCAs) and sugars/sugar anhydrides (S/SAs), were measured in airborne particulate matter in the area of Thessaloniki, northern Greece. The target compounds were measured simultaneously in two particle fractions PM10 and PM2.5 during
The controlled synthesis of polyglucose in one-dimensional coordination nanochannels
Kobayashi Y, et al.
Chemical Communications (Cambridge, England), 52(29), 5156-5159 (2016)
Synthesis of Protected 3-Deoxy-3-fluoro-and 4-Deoxy-4-fluoro-d-galactopyranosides from Levoglucosan
Laine D, et al.
The Journal of Organic Chemistry, 82(9), 4986-4992 (2017)
Bret A Schichtel et al.
Environmental science & technology, 51(17), 9846-9855 (2017-08-02)
Carbonaceous compounds are a significant component of fine particulate matter and haze in national parks and wilderness areas where visibility is protected, i.e., class I areas (CIAs). The Regional Haze Rule set the goal of returning visibility in CIAs on
Jackson Tham et al.
Environmental pollution (Barking, Essex : 1987), 248, 496-505 (2019-03-05)
This study characterizes impacts of peat-forest (PF) smoke on an urban environment through carbonaceous profiles of >260 daily PM2.5 samples collected during 2012, 2013 and 2015. Organic carbon (OC) and elemental carbon (EC) comprising eight carbonaceous fractions are examined for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service