Skip to Content
Merck
All Photos(1)

Key Documents

31140

Sigma-Aldrich

L-(+)-Fructose

≥97.0% (HPLC)

Synonym(s):

L-arabino-2-Hexulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724560
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

solid

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m0/s1

InChI key

BJHIKXHVCXFQLS-FUTKDDECSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Maruyama et al.
Small (Weinheim an der Bergstrasse, Germany), 16(2), e1905916-e1905916 (2019-12-05)
Interest in chiral substances has mainly focused on the substances themselves, but not on the accompanying space, especially regarding the pore alignment. As a method to form both the chiral substance and the accompanying space, cylindrical self-assembly of uniform polystyrene
Jianming Zhang et al.
Probiotics and antimicrobial proteins, 12(1), 32-38 (2019-03-20)
The effects of fructose on plantaricin Q7 biosynthesis in Lactobacillus plantarum (L. plantarum) Q7 were investigated. Fructose induced and accelerated plantaricin Q7 production in the early stages of biosynthesis. Two-dimensional electrophoresis and quantitative real-time reverse transcriptase PCR (qRT-PCR) revealed that
Yi-chun Chen et al.
Chemical senses, 36(3), 237-249 (2011-01-14)
To provide a behavior-based estimate of odor similarity in larval Drosophila, we use 4 recognition-type experiments: 1) We train larvae to associate an odor with food and then test whether they would regard another odor as the same as the
Jun Hu et al.
Applied microbiology and biotechnology, 103(1), 411-425 (2018-11-09)
When sucrose was used as the carbon source, the Basidiomycete Coprinopsis cinerea showed poor growth and low laccase activity in pure culture, but greatly enhanced the level of laccase activity (>1800 U/L) during coculture with the Mucoromycete Gongronella sp. w5. As
Nicolas Rochereau et al.
PLoS biology, 11(9), e1001658-e1001658 (2013-09-27)
Intestinal microfold (M) cells possess a high transcytosis capacity and are able to transport a broad range of materials including particulate antigens, soluble macromolecules, and pathogens from the intestinal lumen to inductive sites of the mucosal immune system. M cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service