Skip to Content
Merck
All Photos(1)

Key Documents

310115

Sigma-Aldrich

Phosphorus trichloride

99.999% trace metals basis

Synonym(s):

Phosphorus(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PCl3
CAS Number:
Molecular Weight:
137.33
Beilstein:
969177
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor density

4.75 (vs air)

Quality Level

vapor pressure

100 mmHg ( 20 °C)
125 mmHg ( 25 °C)
385 mmHg ( 55 °C)

Assay

99.999% trace metals basis

form

liquid

refractive index

n20/D 1.5148 (lit.)

bp

74-78 °C (lit.)

mp

−112 °C (lit.)

density

1.574 g/mL at 20 °C (lit.)

SMILES string

ClP(Cl)Cl

InChI

1S/Cl3P/c1-4(2)3

InChI key

FAIAAWCVCHQXDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Zicane et al.
International journal of peptide and protein research, 48(1), 56-58 (1996-07-01)
N-Formylpyroglutamic acid-7-amido-4-methylcoumarine and pyroglutamyl-pyroglutamic acid-7-amido-4-methylcoumarin are the major products in the synthesis of pyroglutamic acid-7-amido-4-methylcoumarin by phosphorus pentachloride in dimethylformamide and dicyclohexylcarbodiimide under pyridine activation.
Núria Mont et al.
Molecular diversity, 13(1), 39-45 (2008-11-28)
A protocol for the synthesis of N-substituted 2-hydro-4-amino-pyrido[2,3-d]pyrimidin-7(8H)-ones (11) is described. Thus, the formylation of a 2-aminopyridone 12 in 85% formic acid/Ac(2)O, proceeding via in situ cyclization to the intermediate formamide 13, affords the corresponding 2-hydro-4-oxo-pyridopyrimidine 14, which is converted
Action of phosphorus pentachloride on p-sulphonamido benzoic acid.
A D KEMP
Nature, 159(4041), 509-509 (1947-04-12)
Petri A Turhanen et al.
Organic & biomolecular chemistry, 1(18), 3223-3226 (2003-10-07)
Methods for the preparation of mixed tetra-amide esters 1 and 2, the partial amide ester 3, and tri- and P,P-diamides 4 and 5 from monophosphorus spieces 12, 8 and 9, respectively, were developed. Compounds 8 and 9 were obtained from
Phosphorus trichloride-mediated and microwave-assisted synthesis of a small collection of amides bearing strong electron-withdrawing group substituted anilines.
Matteo Colombo et al.
Journal of combinatorial chemistry, 11(3), 335-337 (2009-03-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service