Skip to Content
Merck
All Photos(1)

Key Documents

307556

Sigma-Aldrich

3,3,3-Trifluoro-DL-alanine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(NH2)CO2H
CAS Number:
Molecular Weight:
143.06
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

231-234 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

NC(C(O)=O)C(F)(F)F

InChI

1S/C3H4F3NO2/c4-3(5,6)1(7)2(8)9/h1H,7H2,(H,8,9)

InChI key

HMJQKIDUCWWIBW-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yong Guo et al.
Organic letters, 8(5), 827-829 (2006-02-24)
2-Aminoperfluoropropene has been prepared by the Mg-promoted defluorinative N-silylation of N-p-methoxyphenyl hexafluoroacetone imine and has been employed as a synthon of trifluoroalanine for the preparation of trifluoroalanine dipeptides.
T Clausen et al.
Journal of molecular biology, 262(2), 202-224 (1996-09-20)
Cystathionine beta-lyase (CBL) is a member of the gamma-family of PLP-dependent enzymes, that cleaves C beta-S bonds of a broad variety of substrates. The crystal structure of CBL from E. coli has been solved using MIR phases in combination with
R S Phillips et al.
Archives of biochemistry and biophysics, 296(2), 489-496 (1992-08-01)
Trifluoroalanine is a mechanism-based inactivator of Escherichia coli tryptophan indole-lyase (tryptophanase) and E. coli tryptophan synthase (R. B. Silverman and R. H. Abeles, 1976, Biochemistry 15, 4718-4723). We have found that indole is able to prevent inactivation of tryptophan indole-lyase
C W Fearon et al.
Biochemistry, 21(16), 3790-3794 (1982-08-03)
Inactivation of gamma-cystathionase by beta, beta, beta-trifluoroalanine, a suicide inactivator of the enzyme, results in covalent labeling of an amino group of the protein [Silverman, R. B., & Abeles, R. H. (1977) Biochemistry 16, 5515-5520]. We have established that this
E A Wang et al.
Biochemistry, 20(26), 7539-7546 (1981-12-22)
The alanine racemase from Escherichia coli B has been shown to process DL isomers of beta -fluoroalanine as suicide substrates with an identical partitioning ratio for each enantiomer of 820 catalytic eliminations of HF per enzymatic inactivation event [Wang, E.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service