Skip to Content
Merck
All Photos(3)

Key Documents

303208

Sigma-Aldrich

Bromomethyl acetate

95%

Synonym(s):

Acetoxymethyl bromide, Acetyloxymethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2Br
CAS Number:
Molecular Weight:
152.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

130-133 °C/750 mmHg (lit.)

density

1.56 g/mL at 25 °C (lit.)

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

CC(=O)OCBr

InChI

1S/C3H5BrO2/c1-3(5)6-2-4/h2H2,1H3

InChI key

NHYXMAKLBXBVEO-UHFFFAOYSA-N

General description

The interaction of bromomethyl acetate with O(6)-alkylguanine-DNA alkyltransferase (DNA repair protein) was studied in vitro.

Application

Bromomethyl acetate was used in synthesis of optically active cyclohexene antisepsis agent, ethyl (6R)-6-[N-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate [(R)-1: TAK-242)]. It was also used as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Masami Yamada et al.
Chemical & pharmaceutical bulletin, 54(1), 58-62 (2006-01-06)
Two new synthetic methods were established for the efficient synthesis of optically active cyclohexene antisepsis agent, ethyl (6R)-6-[N-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate [(R)-1: TAK-242)]. The first method involved recrystallization from methanol of the diastereomeric mixture (6RS,1'R)-7, obtained by esterification of carboxylic acid 3 with
A direct preparation of 1, 2-diacetates from aldehydes and ketones promoted by samarium diiodide.
Enholm EJ and Satici H.
Tetrahedron Letters, 32(22), 2433-2436 (1991)
Liping Liu et al.
Chemical research in toxicology, 17(6), 742-752 (2004-06-23)
O(6)-Alkylguanine-DNA alkyltransferase (AGT) is a DNA repair protein that removes O(6)-alkylguanine adducts. The interaction of dibromomethane (CH(2)Br(2)) and bromomethyl acetate (BrCH(2)OAc) with AGT was studied in vitro, and the effect of AGT on their toxicity and mutagenicity was investigated using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service