Skip to Content
Merck
All Photos(2)

Key Documents

300985

Sigma-Aldrich

2,4,6-Trimethoxybenzylamine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH2NH2·HCl
CAS Number:
Molecular Weight:
233.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

204-207 °C (lit.)

SMILES string

Cl.COc1cc(OC)c(CN)c(OC)c1

InChI

1S/C10H15NO3.ClH/c1-12-7-4-9(13-2)8(6-11)10(5-7)14-3;/h4-5H,6,11H2,1-3H3;1H

InChI key

BLFRMOOGAICNSZ-UHFFFAOYSA-N

Application

2,4,6-Trimethoxybenzylamine hydrochloride was used in synthesis of 3-(aminomethyl)isoquinoline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Ohta et al.
Chemical communications (Cambridge, England), (7)(7), 835-837 (2008-02-07)
Copper(i)-catalysed domino four-component coupling-cyclisation using 2-ethynylbenzaldehydes, paraformaldehyde, secondary amine, and t-BuNH(2) in DMF leads to direct and efficient formation of 3-(aminomethyl)isoquinolines in good to high yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service