Skip to Content
Merck
All Photos(1)

Key Documents

300764

Sigma-Aldrich

1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole

98%

Synonym(s):

Noreleagnine, THBC, Tetrahydronorharman, Tryptoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2
CAS Number:
Molecular Weight:
172.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

mp

206-208 °C (lit.)

SMILES string

C1Cc2c(CN1)[nH]c3ccccc23

InChI

1S/C11H12N2/c1-2-4-10-8(3-1)9-5-6-12-7-11(9)13-10/h1-4,12-13H,5-7H2

InChI key

CFTOTSJVQRFXOF-UHFFFAOYSA-N

Gene Information

General description

Ozonolysis of the enamine bond of 1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole derivatives was studied.

Application

1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole was employed in alkaloid synthesis and in studies on neurodegenerative diseases.
  • Reactant for synthesis of the indolyl-β-carboline alkaloid eudistomin U via IBX mediated room temperature oxidative aromatization
  • Reactant for preparation of neuroprotective HDAC6 inhibitors
  • Reactant for preparation of aminofuranopyrimidines as EGFR and Aurora A kinase inhibitors
  • Reactant for preparation of inhibitors of CDK4
  • Reactant for preparation of tetrahydrocarboline derivatives of as human 5-HT5A receptor ligands
  • Reactant for preparation of 5-(diaminomethyl)-2,4-aminopyrimidines as dihydrofolate reductase inhibitors and antibacterial agents

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang-Yuan Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(7), 2478-2483 (2013-01-11)
Natural product discovery by random screening of broth extracts derived from cultured bacteria often suffers from high rates of redundant isolation, making it ever more challenging to identify novel biologically interesting natural products. Here we show that homology-based screening of
I M McDonald et al.
Journal of medicinal chemistry, 43(19), 3518-3529 (2000-09-23)
A novel series of nonpeptide CCK(2) receptor antagonists has been prepared, in which 2,7-dioxo-2,3,4,5,6,7-hexahydro-1H-benzo[h][1, 4]diazonine (5) was used as a chemical template. This uncommon ring system was obtained in a highly substituted form and in high yield by ozonolysis of
Chem. Abstr., 121, 54854s-54854s (1994)
J. Chin. Chem. Soc., 41, 231-231 (1994)
Christian O Alberto et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(29), 10671-10676 (2011-07-22)
Melanin-concentrating hormone (MCH) is a hypothalamic neuropeptide that promotes positive energy balance and anxiety. Since dopamine (DA) is also closely implicated in these functions, the present study investigated the effect of DA on MCH neurons. Using whole-cell patch-clamp recordings in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service