Skip to Content
Merck
All Photos(1)

Key Documents

297666

Sigma-Aldrich

2-Isobutyl-3-methoxypyrazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.49 (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

COc1nccnc1CC(C)C

InChI

1S/C9H14N2O/c1-7(2)6-8-9(12-3)11-5-4-10-8/h4-5,7H,6H2,1-3H3

InChI key

UXFSPRAGHGMRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Isobutyl-3-methoxypyrazine, a potent bell-pepper odourant, has been found to bind to cow olfactory mucosa homogenate.

Application

2-Isobutyl-3-methoxypyrazine has been employed as a model odorant molecule during an electrochemical assay for the detection of odorant molecules based on a rat odorant-binding protein (rOBP3).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilie Barou et al.
Bioelectrochemistry (Amsterdam, Netherlands), 101, 28-34 (2014-07-18)
We developed an electrochemical assay for the detection of odorant molecules based on a rat odorant-binding protein (rOBP3). We demonstrated that rOBP3 cavity binds 2-methyl-1,4-naphtoquinone (MNQ), an electrochemical probe, as depicted from the decrease of its electrochemical signal, and deduced
P Pelosi et al.
The Biochemical journal, 201(1), 245-248 (1982-01-01)
2-Isobutyl-3-methoxypyrazine, a potent bell-pepper odorant, binds to cow olfactory mucosa homogenate. The receptor is saturable in the micromolar range and is competitively inhibited by other bell-pepper odourants, but not by other pyrazines of different odours. Other tissues do not bind
Imelda Ryona et al.
Journal of agricultural and food chemistry, 56(22), 10838-10846 (2008-10-24)
The effects of light exposure on 3-isobutyl-2-methoxypyrazine (IBMP) accumulation and degradation in Vitis vinifera L. cv. Cabernet Franc berries were assessed by comparison of shaded and exposed clusters within the same vine throughout a growing season. Twenty-seven vines were shoot-thinned
Leila Denise Falcão et al.
Journal of agricultural and food chemistry, 55(9), 3605-3612 (2007-03-31)
This report has investigated the seasonal temperatures influences (winter and summer) of five vineyards at different altitudes on the concentrations of 2-methoxy-3-isobutylpyrazine (MIBP), alpha- and beta-ionone, and beta-damascenone in 2004 or 2005 vintages of Cabernet Sauvignon wines from Santa Catarina
Maria Staiano et al.
Biopolymers, 89(4), 284-291 (2007-12-22)
The effects of hydrostatic pressure on the structure and stability of porcine odorant-binding protein (pOBP) in the presence and absence of the odorant molecule 2-isobutyl-3-methoxypyrazine (IBMP) were studied by steady-state and time-resolved fluorescence spectroscopy as well as by molecular dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service