Skip to Content
Merck
All Photos(1)

Key Documents

294543

Sigma-Aldrich

2-(2-Thiazolylazo)-p-cresol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N3OS
CAS Number:
Molecular Weight:
219.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

130-132 °C (lit.)

solubility

soluble 50 mg/mL, clear, red to red-brown (ethanol:CHCl3 (1:1))

SMILES string

Cc1ccc(O)c(c1)\N=N\c2nccs2

InChI

1S/C10H9N3OS/c1-7-2-3-9(14)8(6-7)12-13-10-11-4-5-15-10/h2-6,14H,1H3/b13-12+

InChI key

MZRKINSTWYZJLV-OUKQBFOZSA-N

Application

2-(2-Thiazolylazo)-p-cresol is a heterocyclic azo-dye2, has been used:
  • as iron binding ligand during determination of iron speciation in seawater by cathodic stripping voltammetry
  • in spectrophotometric determination of vanadium, titanium, lead, indium and nickel
  • as as a spectrophotometric reagent in the determination of uranium in mine drainage water samples

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of uranium using 2-(2-thiazolylazo)-p-cresol (TAC) in the presence of surfactants.
Teixeira LSG, et al.
Journal of the Brazilian Chemical Society, 10(6), 519-522 (1999)
Determination of Iron Speciation by Cathodic Stripping Voltammetry in Seawater Using the Competing Ligand 2-(2-Thiazolylazo)-p-cresol (TAC).
Croot PL and Johansson M.
Electroanalysis, 12(8), 565-576 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service