Skip to Content
Merck
All Photos(1)

Key Documents

292206

Sigma-Aldrich

2-Methyl-1,3-dioxolane

97%

Synonym(s):

Acetaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102520
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.398 (lit.)

bp

82-83 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

CC1OCCO1

InChI

1S/C4H8O2/c1-4-5-2-3-6-4/h4H,2-3H2,1H3

InChI key

HTWIZMNMTWYQRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The kinetics and mechanism of the gas-phase thermal decomposition of 2-methyl-1,3-dioxolane has been studied in a static system. The infrared spectra of solid, liquid and gaseous 2-methyl-1,3-dioxolane has been studied. Low-temperature ozonation of 2-methyl-1,3-dioxolane in acetone-d6, methyl acetate and tert-butyl methyl ether has been reported.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E T Iwamoto et al.
The Journal of pharmacology and experimental therapeutics, 266(1), 329-338 (1993-07-01)
The present study was designed to characterize the antinociception produced by the administration of a potent muscarinic agonist into the intrathecal space of the lumbar spinal cord of male Sprague-Dawley rats. Seven days after surgical implantation of intrathecal catheters, animals
A Closse et al.
Naunyn-Schmiedeberg's archives of pharmacology, 335(4), 372-377 (1987-04-01)
Special conditions--tricine buffer containing Ca2+ and Mg2+, 22 degrees C (TCM)--allow to label a much higher proportion of muscarinic receptors by [3H]cis-methyldioxolane (CD) than hitherto described (Vickroy et al. 1984a). Taking the maximum number of binding sites, Bmax, of [3H]QNB
H I Yamamura et al.
Brain research, 325(1-2), 340-344 (1985-01-28)
(+)-cis-[3H]Methyldioxolane ((+)-[3H]CD), a potent muscarinic agonist, was used to label high-affinity agonist states of muscarinic receptors in thin tissue sections of the rat central nervous system. Light microscopic autoradiography of atropine-sensitive (+)-[3H]CD binding sites revealed regions of dense labeling (superior
R A Huff et al.
Journal of neurochemistry, 62(1), 388-391 (1994-01-01)
cis-Methyldioxolane (CD) is a muscarinic receptor agonist. [3H]CD has been used to label a subpopulation of muscarinic receptors described as exhibiting high agonist affinity. Pharmacological evidence suggests that the population of receptors labeled by [3H]CD consists of m2 and/or m4
M Watson et al.
Brain research, 346(2), 387-391 (1985-11-04)
The relationship of choline acetyltransferase (ChAT) activity and high affinity binding of the potent and selective sodium-dependent choline uptake inhibitor [3H]hemicholinium-3 ([3H]HC-3) to high-affinity binding of the muscarinic agonist [3H](+)-cis-methyldioxolane ([3H](+)CD), the putative M1 selective antagonist [3H]pirenzepine ([3H]PZ) and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service