Skip to Content
Merck
All Photos(2)

Key Documents

284572

Sigma-Aldrich

4-Aminobenzamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CONH2
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

181-183 °C (lit.)

functional group

amide

SMILES string

NC(=O)c1ccc(N)cc1

InChI

1S/C7H8N2O/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H2,9,10)

InChI key

QIKYZXDTTPVVAC-UHFFFAOYSA-N

Related Categories

Application

4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Burgman et al.
Radiation research, 116(3), 406-415 (1988-12-01)
The purpose of this study was to investigate a possible involvement of poly(ADP-ribosyl)ation reactions in hyperthermic cell killing and hyperthermic DNA strand-break induction and repair in HeLa S3 cells. The inhibitors of poly(ADP-ribose) polymerase, 3-aminobenzamide (3AB) and 4-aminobenzamide (4AB), were
R Laffranchi et al.
Experimental cell research, 237(1), 217-222 (1998-01-07)
Interferon-gamma is among the cytokines which have been implicated as effector molecules of beta-cell destruction in autoimmune diabetes. Its mechanism of action is, however, largely unknown. In the present study rat pancreatic beta-cells, INS-1, were incubated with rat interferon-gamma (rIRN-gamma)
Jamie Lajiness et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 5(3), 216-226 (2009-05-16)
Imidazole and pyrrole-containing polyamides belong to an important class of compounds that can be designed to target specific DNA sequences, and they are potentially useful in applications of controlling gene expression. The extent of polyamide curvature is an important consideration
E Ben-Hur et al.
Radiation research, 97(3), 546-555 (1984-03-01)
Postirradiation incubation of V79 Chinese hamster cells with inhibitors of poly(ADP-ribose) synthesis was found to potentiate the killing of cells by X rays. Potentiation increased with incubation time and with concentration of the inhibitor. Preirradiation incubation had only a small
D Monti et al.
Biochemical and biophysical research communications, 199(2), 525-530 (1994-03-15)
The death of target cells by cytotoxic effector cells is a relevant biological phenomenon, where cells are activated and a very quick apoptotic program occurs. In order to test the hypothesis that the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP) plays a role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service