Skip to Content
Merck
All Photos(2)

Key Documents

283207

Sigma-Aldrich

Magnesium bis(monoperoxyphthalate) hexahydrate

greener alternative

80%, technical grade

Synonym(s):

MMPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CC6H4CO3)2Mg·6H2O
Molecular Weight:
494.64
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

93 °C (dec.) (lit.)

functional group

hydroperoxide

greener alternative category

SMILES string

O.O.O.O.O.O.OOC(=O)c1ccccc1C(=O)O[Mg]OC(=O)c2ccccc2C(=O)OO

InChI

1S/2C8H6O5.Mg.6H2O/c2*9-7(10)5-3-1-2-4-6(5)8(11)13-12;;;;;;;/h2*1-4,12H,(H,9,10);;6*1H2/q;;+2;;;;;;/p-2

InChI key

WWOYCMCZTZTIGU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product MMPP is an alternative to widely used oxidant MCPBA as it is cheap, stable in the solid state, safer in handling and easier to use and thus aligns with the principle "Designing Safer Chemicals". For more information see the below articles.
Magnesium bis(monoperoxyphthalate) hexahydrate as mild and efficient oxidant for the synthesis of selenones.
Magnesium Bis(monoperoxyphthalate) Hexahydrate (MMPP).
Highly efficient epoxidation of unsaturated steroids using magnesium bis(monoperoxyphthalate) hexahydrate.

Application

A safer, greener oxidizing reagent that is a suitable replacement for peroxycarboxylic acids and is capable of effecting many types of oxidation reactions.

Oxidation Reactions Using Magnesium Monoperphthalate: A Comparison with m-Chloroperoxybenzoic Acid
Reagent for:
  • Trans-diaxial hydroxylation under bismuth(III) triflate catalyst
  • Oxidation rections under mild conditions
  • Epoxidation of unsaturated steroids
  • Synthesis of ring-B oxygenated steroids as anticancer agents

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Org. Perox. E - Skin Corr. 1C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Luo et al.
Lipids in health and disease, 16(1), 182-182 (2017-09-28)
Estrogen was reported to protect against obesity, however the mechanism remains unclear. We aimed to investigate the impact of 17β-estradiol (17β-E2) on triglyceride metabolism in adipocytes with or without lipopolysacchride (LPS) stimulating, providing novel potential mechanism for estrogen action. 3T3-L1
Wasserman, H. H. et al
Tetrahedron Letters, 43, 3351-3351 (2002)
Chan-Woo Park et al.
Journal of microbiology and biotechnology, 27(7), 1359-1366 (2017-05-18)
(
Heaney, H.; Newbold, A. J.
Tetrahedron Letters, 42, 6607-6607 (2001)
Ming-Yi Chen et al.
The Journal of organic chemistry, 69(8), 2884-2887 (2004-04-13)
A protocol that uses moist magnesium monoperoxyphthalate (MMPP) as an oxidant under microwave irradiation rapidly yields a variety of glycosyl sulfoxides from corresponding sulfides in high yields with high selectivity.

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service