278505
Sulfuryl chloride solution
1.0 M in methylene chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
SO2Cl2
CAS Number:
Molecular Weight:
134.97
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
form:
liquid
Recommended Products
form
liquid
Quality Level
concentration
1.0 M in methylene chloride
density
1.352 g/mL at 25 °C
SMILES string
ClS(Cl)(=O)=O
InChI
1S/Cl2O2S/c1-5(2,3)4
InChI key
YBBRCQOCSYXUOC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Palladium-Catalyzed Synthesis of Ammonium Sulfinates from Aryl Halides and a Sulfur Dioxide Surrogate: A Gas-and Reductant-Free Process: This study elaborates on using sulfuryl chloride for oxidative chlorination leading to sulfonyl chloride formation, which is important for synthetic organic chemistry (Emmett et al., 2014).
- Combining Organometallic Reagents, the Sulfur Dioxide Surrogate DABSO, and Amines: A One‐Pot Preparation of Sulfonamides, Amenable to Array Synthesis: This research discusses an innovative one-pot method for preparing sulfonamides using sulfuryl chloride, highlighting its utility in streamlining complex syntheses (Deeming et al., 2015).
- Interfacial polymerization: from chemistry to functional materials: The article illustrates the use of sulfuryl chloride in interfacial polymerization, a key technique for developing advanced materials with tailored properties (Zhang et al., 2020).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Carc. 2 - Eye Dam. 1 - Skin Corr. 1C - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Qian Wu et al.
Chemical communications (Cambridge, England), 47(32), 9188-9190 (2011-07-14)
A new, efficient protocol for the synthesis of di(hetero)aryl sulfides is described. Cheap and easily available arylsulfonyl chlorides as a sulfur source reductively couple with electron-rich (hetero)arenes (e.g., indolizines, indoles, electron-rich benzenes, etc.) in the presence of triphenylphosphine to afford
Philippe Hermange et al.
Organic letters, 11(18), 4044-4047 (2009-08-15)
Reaction of an isoquinoline, a silyloxyfuran, and an acyl or sulfonyl chloride provides easy access to a wide variety of isoquinolinobutyrolactones with excellent yields and diastereoselectivites (R*,R* isomer), even in the case of formation of quaternary centers (i.e., R(3) or
Ourida Saidi et al.
Journal of the American Chemical Society, 133(48), 19298-19301 (2011-11-04)
A selective catalytic meta sulfonation of 2-phenylpyridines was found to occur in the presence of (arene)ruthenium(II) complexes upon reaction with sulfonyl chlorides. The 2-pyridyl group facilitates the formation of a stable Ru-C(aryl) σ bond that induces a strong para-directing effect.
Tandem cyclizations of 1,6-enynes with arylsulfonyl chlorides by using visible-light photoredox catalysis.
Guo-Bo Deng et al.
Angewandte Chemie (International ed. in English), 52(5), 1535-1538 (2013-01-05)
Xiaoming Zeng et al.
Journal of the American Chemical Society, 133(44), 17638-17640 (2011-10-13)
A variety of polysubstituted 1H-indenes can be prepared through the copper-catalyzed arylative cyclization of simple arylalkynes with commercially available aromatic sulfonyl chlorides that function as an aryl group donor. The reaction tolerates a broad range of functional groups, including bromide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service